missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, HPLC Grade, ≥ 99.8%, LabChem™

Supplier:  LabChem LC168104

 View more versions of this product

Catalog No. LC168104


Explore available promotions

Description

Description

Specifications

Specifications

Methanol
Passes Test
100
≤0.0002meq/g
Passes Test
Liquid
CH4O
MFCD00004595
≤0.1%
Soluble in water
CO
32.04
CHEBI:17790
≥99.8%
67-56-1
Colorless
≤0.0003meq/g
4 L
Amber Glass
≤0.001%
CH3OH
UN1230
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
HPLC

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (liver, kidneys, central nervous system, optic nerve) (dermal, oral).

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.

Danger

EINECSNumber : 200-659-6

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions