Learn More
Methanol, Optima™ for HPLC, Fisher Chemical™
High purity mobile phase for HPLC
$183.63 - $531.67
Chemical Identifiers
| CAS | 67-56-1 |
|---|---|
| Molecular Formula | CH4O |
| Molecular Weight (g/mol) | 32.04 |
| MDL Number | MFCD00004595 |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
| PubChem CID | 887 |
| ChEBI | CHEBI:17790 |
| IUPAC Name | methanol |
| SMILES | CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
A454-1
|
Thermo Fisher Scientific
BPA4541 |
1 L | Wide Neck Amber Bottle |
Each for $183.63
Case of 6 Each for $1,101.78
|
|
||||
|
A454-4
|
Fisher Chemical
BPA4544 |
4 L | Amber Soda Lime Glass |
Each for $321.08
Case of 4 Each for $1,284.32
|
|
||||
|
A454SK-4
|
Thermo Fisher Scientific
A454SK4 |
4 L | Safe-Cote™ Amber Glass |
Each for $531.67
Case of 4 Each for $2,126.68
|
|
||||
Description
High purity mobile phase for HPLC
- Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
- Tested for HPLC gradient suitability
- Mobile phase for analytical and preparative separations
- Also meets ACS specifications
- Intended for research use only
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Specifications
| -98°C | |
| 0.01 AU max. at 254nm, 0.11 AU max. at 230nm, 0.23 AU max. at 220nm, 1.00 AU max. at 205nm | |
| 0.791g/cm³ | |
| 1 L | |
| Pass Test | |
| Liquid | |
| 99.9% min by GC | |
| CH4O | |
| CH3OH | |
| 128 hPa at 20°C | |
| 1230 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| ≥99.9% | |
| Methanol |
| 67-56-1 | |
| Colorless | |
| 64.7°C | |
| 12°C (53.6°F) | |
| Wide Neck Amber Bottle | |
| 1ppm max. | |
| 0.0003mEq/g max. | |
| 0.0002mEq/g max. | |
| MFCD00004595 | |
| 0.55 cP at 20°C | |
| 0.07% max. | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04 | |
| Also meets ACS specifications |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.
NFPA
Health:1
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid