missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™

Supplier:  Fisher BioReagents FLBP11054

 View more versions of this product

Catalog No. BP1105-4



Explore available promotions

Description

Description

Methanol (Peroxide-Free/Sequencing)

Methanol is used in staining and destaining protein electrophoresis gels, in HPLC, and in other biological applications.
Specifications

Specifications

Methanol
Peroxide-free
0.025 max. at 254nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 1 max. at 205nm
0.791g/cm³
4 L
Liquid
1.326 to 1.330 (at 25°C)
≥99.9 %
CH4O
CH3OH
128 hPa at 20°C
0.1% max.
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.04
CHEBI:17790
≥99.9%
-98°C
67-56-1
Colorless
64.7°C
Glass Bottle
Peroxides: ≤0001%
3ppm max.
0.0003mEq/g max.
0.0002mEq/g max.
MFCD00004595
0.55 cP at 20°C
15, 6029
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
Sequencing

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

Recommended Storage : RT

SDS Documents

Documents

Accessories
Promotions

Promotions