missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol Purified, Fisherbrand™, Histoprep™ Edge
Edge

Supplier:  Epredia HC4001GAL

Catalog No. HC-400-1GAL



Edge
Explore available promotions

Description

Description

Specifications

Specifications

Liquid
-98°C
67-56-1
Poly Bottle
0.55 cP at 20°C
1 gal.
CH4O
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.04
CHEBI:17790
Methanol Purified
64.7°C
0.791g/cm³
128 hPa at 20°C
Colorless
Assay: >95%
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
Videos
SDS
Documents

Documents

Promotions

Promotions