missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, UHPLC-MS, Thermo Scientific™

Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer.

Supplier:  Thermo Fisher Scientific A4581

Catalog No. A4581



Explore available promotions

Description

Description

Methanol, CH4O, CAS Number-67-56-1, wood naphtha, pyroxylic spirit, methyl alcohol, methylol, columbian spirit, colonial spirit, wood spirit, wood alcohol, methyl hydroxide, carbinol, 1L

Specifications

Specifications

Methanol
67-56-1
Colorless
64.7°C
Clear Borosilicate Glass Bottle
1ppm max.
0.0003meq/g max.
0.0002meq/g max.
MFCD00004595
0.55 cP at 20°C
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.04
CHEBI:17790
-98°C
≤0.005 AU at 280nm, ≤0.01 AU at 254nm, ≤0.1 AU at 230nm, ≤0.4 AU at 215nm, ≤000 5 AU at 260nm, ≤02. AU at 220nm, ≤05. AU at 210nm
0.791 g/cm3
1 L
Liquid
99.9% minimum by GC
CH4O
CH3OH
128 hPa at 20°C
0.02% Max.
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
UHPLC-MS
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions

RUO – Research Use Only