Thermo Scientific™ Methoxamine (MOX) Reagent
Prepare oximes of steroids and ketoacids prior to silylation with the Thermo Scientific™ Methoxamine (MOX) Reagent.
Supplier:  Thermo Scientific™ TS45950
Catalog No. PI45950
Explore available promotions
Description
Description
Prepare oximes of steroids and ketoacids prior to silylation with the Thermo Scientific™ Methoxamine (MOX) Reagent. It prevents the formation of multiple derivatives when enols are present during silylation, thereby ensuring optimal quantitation. The reagent is a 2% solution of methoxyamine–hydrogen chloride in pyridine.
  • 2% methoxyamine·HCl (M.W. 83.51) in pyridine
  • Prevents formation of multiple derivatives when enols are present during silylation
  • Supplied in amber Thermo Scientific Hypo-Vial Sample Storage Vial with septum and crimp top
Specifications
Specifications
Preparing Oximes of Steroids and Ketoacids
Corrosive, Flammable
2°C to 8°C
95 - 98, 1 - 3%
Hypo-Vial Container; 10 mL
Pyridine, Methoxyammonium chloride
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.