missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Methoxycarbonylmethylene)triphenylphosphorane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1402022

 View more versions of this product

Catalog No. AAA1402022


Explore available promotions

Description

Description

(Methoxycarbonylmethylene)triphenylphosphorane is used in olefination reactions. Further, it undergoes the Wittig reaction with aldehydes to give substituted methyl acrylates. It is used in the preparation of (triphenylphosphoranylidene)-ketene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Methoxycarbonylmethylene)triphenylphosphorane is used in olefination reactions. Further, it undergoes the Wittig reaction with aldehydes to give substituted methyl acrylates. It is used in the preparation of (triphenylphosphoranylidene)-ketene.

Solubility
Soluble in chloroform. Slightly soluble in ethanol and terahydrofuran. Insoluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

(Methoxycarbonylmethylene)triphenylphosphorane
165°C to 169°C
MFCD00008455
618430
methyl triphenylphosphoranylidene acetate, carbomethoxymethylene triphenylphosphorane, methyl 2-triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, methyl ester, methyl triphenylphosphoranylideneacetate, triphenylphosphoranylidene acetic acid methyl ester, triphenyl carbomethoxymethylene phosphorane, methoxycarbonylmethylene triphenylphosphorane, acetic acid, 2-triphenylphosphoranylidene-, methyl ester
NTNUDYROPUKXNA-UHFFFAOYSA-N
methyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
17453
98%
2605-67-6
C21H19O2P
100 g
Air Sensitive
Soluble in chloroform. Slightly soluble in ethanol and terahydrofuran. Insoluble in water.
COC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
334.36
334.36

Chemical Identifiers

2605-67-6
334.36
NTNUDYROPUKXNA-UHFFFAOYSA-N
17453
COC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C21H19O2P
MFCD00008455
methyl triphenylphosphoranylidene acetate, carbomethoxymethylene triphenylphosphorane, methyl 2-triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, methyl ester, methyl triphenylphosphoranylideneacetate, triphenylphosphoranylidene acetic acid methyl ester, triphenyl carbomethoxymethylene phosphorane, methoxycarbonylmethylene triphenylphosphorane, acetic acid, 2-triphenylphosphoranylidene-, methyl ester
methyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 220-018-4

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only