Learn More
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 210490050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
Methoxylamine hydrochloride | |
148°C to 153°C | |
Authentic | |
Glass bottle | |
CH3ONH2·HCl | |
5 g | |
01,670 | |
o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride | |
XNXVOSBNFZWHBV-UHFFFAOYSA-N | |
O-methylhydroxylamine;hydrochloride | |
521874 | |
98+% |
593-56-6 | |
White to Yellow | |
98+% | |
CH5NO·HCl | |
MFCD00012951 | |
01, 288 | |
15, 6060 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
CON.Cl | |
83.52 | |
83.52 | |
Crystalline Powder |
Chemical Identifiers
593-56-6 | |
83.52 | |
XNXVOSBNFZWHBV-UHFFFAOYSA-N | |
521874 | |
CON.Cl |
CH5NO·HCl | |
MFCD00012951 | |
o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride | |
O-methylhydroxylamine;hydrochloride |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 209-798-7
RUO – Research Use Only