Learn More
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
$118.70 - $403.59
Chemical Identifiers
CAS | 593-56-6 |
---|---|
Molecular Formula | CH6ClNO |
Molecular Weight (g/mol) | 83.515 |
MDL Number | MFCD00012951 |
InChI Key | XNXVOSBNFZWHBV-UHFFFAOYSA-N |
Synonym | o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride |
PubChem CID | 521874 |
IUPAC Name | O-methylhydroxylamine;hydrochloride |
SMILES | CON.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1918806
|
Thermo Scientific Chemicals
A1918806 |
5 g |
Each for $118.70
|
|
||||||
AAA1918814
|
Thermo Scientific Chemicals
A1918814 |
25 g |
Each for $403.59
|
|
||||||
Description
Methoxylamine hydrochloride is used as a reagent in the preparation of O-methyl oximes from aldehydes. It is orally bioavailable small molecule inhibitor with potential adjuvant activity. It is also used as internal standard for prostaglandin assays by gas chromatography-mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethoxylamine hydrochloride is used as a reagent in the preparation of O-methyl oximes from aldehydes. It is orally bioavailable small molecule inhibitor with potential adjuvant activity. It is also used as internal standard for prostaglandin assays by gas chromatography-mass spectrometry.
Solubility
Soluble in water, dimethyl sulfoxide and methanol.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and acid anhydrides.
Chemical Identifiers
593-56-6 | |
83.515 | |
XNXVOSBNFZWHBV-UHFFFAOYSA-N | |
521874 | |
CON.Cl |
CH6ClNO | |
MFCD00012951 | |
o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride | |
O-methylhydroxylamine;hydrochloride |
Specifications
593-56-6 | |
CH6ClNO | |
5 g | |
3589723 | |
14,5989 | |
Soluble in water,dimethyl sulfoxide and methanol. | |
CON.Cl | |
83.515 | |
83.52 | |
Methoxylamine hydrochloride |
∼148°C (decomposition) | |
MFCD00012951 | |
UN3261 | |
Hygroscopic | |
o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride | |
XNXVOSBNFZWHBV-UHFFFAOYSA-N | |
O-methylhydroxylamine;hydrochloride | |
521874 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H314-H318-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 209-798-7
RTECSNumber : NC3980000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only