Learn More
(Methoxymethyl)triphenylphosphonium chloride, 98+%, Thermo Scientific Chemicals
$112.14 - $891.18
Chemical Identifiers
CAS | 4009-98-7 |
---|---|
Molecular Formula | C20H20ClOP |
Molecular Weight (g/mol) | 342.80 |
MDL Number | MFCD00011800 |
InChI Key | SJFNDMHZXCUXSA-UHFFFAOYSA-M |
Synonym | methoxymethyl triphenylphosphonium chloride, methoxymethyl triphenylphosphanium chloride, phosphonium, methoxymethyl triphenyl-, chloride, methoxymethyltriphenylphosphonium chloride, methoxymethyl-triphenylphosphonium chloride, methoxymethyl-triphenyl-phosphonium chloride, triphenyl methoxymethyl phosphonium chloride, methoxy methyl triphenyl phosphonium chloride |
PubChem CID | 2723798 |
IUPAC Name | methoxymethyl(triphenyl)phosphanium;chloride |
SMILES | [Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1438014
|
Thermo Scientific Chemicals
A1438014 |
25 g |
N/A
|
|
||||||
AAA1438022
|
Thermo Scientific Chemicals
A1438022 |
100 g |
N/A
|
|
||||||
AAA1438036
|
Thermo Scientific Chemicals
A1438036 |
500 g |
N/A
|
|
||||||
Description
(Methoxymethyl)triphenylphosphonium chloride is used as a phase transfer catalyst and in the synthesis of taxol-A fragment. It is widely used in the synthesis of pharmaceutical product cephalotaxine, which is used as an antiviral and antitumor agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(Methoxymethyl)triphenylphosphonium chloride is used as a phase transfer catalyst and in the synthesis of taxol-A fragment. It is widely used in the synthesis of pharmaceutical product cephalotaxine, which is used as an antiviral and antitumor agent.
Solubility
Soluble in methanol and chloroform. It decomposes in water.
Notes
Hygroscopic . Incompatible with strong oxidizing agents.
Chemical Identifiers
4009-98-7 | |
342.80 | |
SJFNDMHZXCUXSA-UHFFFAOYSA-M | |
2723798 | |
[Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C20H20ClOP | |
MFCD00011800 | |
methoxymethyl triphenylphosphonium chloride, methoxymethyl triphenylphosphanium chloride, phosphonium, methoxymethyl triphenyl-, chloride, methoxymethyltriphenylphosphonium chloride, methoxymethyl-triphenylphosphonium chloride, methoxymethyl-triphenyl-phosphonium chloride, triphenyl methoxymethyl phosphonium chloride, methoxy methyl triphenyl phosphonium chloride | |
methoxymethyl(triphenyl)phosphanium;chloride |
Specifications
4009-98-7 | |
C20H20ClOP | |
25 g | |
Hygroscopic | |
Soluble in methanol and chloroform. It decomposes in water. | |
[Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
342.80 | |
342.81 | |
(Methoxymethyl)triphenylphosphonium chloride |
∼190°C (decomposition) | |
MFCD00011800 | |
924215 | |
methoxymethyl triphenylphosphonium chloride, methoxymethyl triphenylphosphanium chloride, phosphonium, methoxymethyl triphenyl-, chloride, methoxymethyltriphenylphosphonium chloride, methoxymethyl-triphenylphosphonium chloride, methoxymethyl-triphenyl-phosphonium chloride, triphenyl methoxymethyl phosphonium chloride, methoxy methyl triphenyl phosphonium chloride | |
SJFNDMHZXCUXSA-UHFFFAOYSA-M | |
methoxymethyl(triphenyl)phosphanium;chloride | |
2723798 | |
≥98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 223-664-5
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only