missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-hydroxy-1-cyclopropane carboxylate, 90%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 302110050

 View more versions of this product

Catalog No. AC302110050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl 1-hydroxy-1-cyclopropane carboxylate
Yellow
17°C
88% min. (GC)
C5H8O3
MFCD02093884
methyl 1-hydroxycyclopropanecarboxylate, methyl 1-hydroxy-1-cyclopropane carboxylate, methyl 1-hydroxy-1-cyclopropanecarboxylate, 1-hydroxycyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropanecarboxylic acid methyl ester, methyl-1-hydroxy-1-cyclopropane carboxylate, hydroxy-cyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropane-1-carboxylic acid methyl ester, cyclopropanecarboxylic acid, 1-hydroxy-, methyl ester
KPJWVJURYXOHOO-UHFFFAOYSA-N
methyl 1-hydroxycyclopropane-1-carboxylate
2733178
90%
33689-29-1
70°C (13.0 mmHg)
Authentic
Glass bottle
1.4470 to 1.452
5 g
Solubility in water: .. Other solubilities: soluble in most organic solvents
COC(=O)C1(O)CC1
116.12
116
Liquid

Chemical Identifiers

33689-29-1
116.12
KPJWVJURYXOHOO-UHFFFAOYSA-N
2733178
COC(=O)C1(O)CC1
C5H8O3
MFCD02093884
methyl 1-hydroxycyclopropanecarboxylate, methyl 1-hydroxy-1-cyclopropane carboxylate, methyl 1-hydroxy-1-cyclopropanecarboxylate, 1-hydroxycyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropanecarboxylic acid methyl ester, methyl-1-hydroxy-1-cyclopropane carboxylate, hydroxy-cyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropane-1-carboxylic acid methyl ester, cyclopropanecarboxylic acid, 1-hydroxy-, methyl ester
methyl 1-hydroxycyclopropane-1-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only