missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5274506

 View more versions of this product

Catalog No. AAH5274506


Explore available promotions

Description

Description

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate is a useful reagent for the trifluoromethylation of alkyl halides. It is also broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate is a useful reagent for the trifluoromethylation of alkyl halides. It is also broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications

Specifications

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate
1.509
47°C (117°F)
1.351
MFCD00144316
UN2920
methyl 2,2-difluoro-2-fluorosulfonyl acetate, methylfluorosulphonyldifluoroacetate, methyl difluoro fluorosulfonyl acetate, methyl fluorosulfonyldifluoroacetate, methyl fluorosulphonyl difluoro acetate, methyl fluorosulphonyldifluoroacetate, methyl 2-fluorosulfonyl difluoroacetate, fluorosulfonyldifluoroacetic acid methyl ester, 2,2-difluoro-2-fluorosulfonylacetic acid methyl ester, difluoro fluorosulfonyl acetic acid methyl ester
COC(=O)C(F)(F)S(=O)(=O)F
192.108
192.11
680-15-9
117°C to 118°C
C3H3F3O4S
FSO2CF2CO2CH3
5 g
1812896
GQJCAQADCPTHKN-UHFFFAOYSA-N
methyl 2,2-difluoro-2-fluorosulfonylacetate
547667
97%

Chemical Identifiers

680-15-9
192.108
GQJCAQADCPTHKN-UHFFFAOYSA-N
547667
COC(=O)C(F)(F)S(=O)(=O)F
C3H3F3O4S
MFCD00144316
methyl 2,2-difluoro-2-fluorosulfonyl acetate, methylfluorosulphonyldifluoroacetate, methyl difluoro fluorosulfonyl acetate, methyl fluorosulfonyldifluoroacetate, methyl fluorosulphonyl difluoro acetate, methyl fluorosulphonyldifluoroacetate, methyl 2-fluorosulfonyl difluoroacetate, fluorosulfonyldifluoroacetic acid methyl ester, 2,2-difluoro-2-fluorosulfonylacetic acid methyl ester, difluoro fluorosulfonyl acetic acid methyl ester
methyl 2,2-difluoro-2-fluorosulfonylacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only