missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 2,2-dimethyl-3-hydroxypropionate, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 210911000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl 2,2-dimethyl-3-hydroxypropionate | |
1.0400g/mL | |
76°C | |
98% min. (GC) | |
C6H12O3 | |
MFCD00009707 | |
03, 331 | |
hydroxypivalic acid methyl ester, methyl 3-hydroxypivalate, methyl hydroxypivalate, methyl 2,2-dimethyl-3-hydroxypropionate, hydroxypivalicacidmethylester, propanoic acid, 3-hydroxy-2,2-dimethyl-, methyl ester, 2,2-dimethyl-3-hydroxypropionic acid methyl ester, 3-hydroxy-2,2-dimethyl-propionic acid methyl ester, methyl 2,2-dimethyl-3-hydroxypropanoate, methyl hydroxylpivalate | |
COC(=O)C(C)(C)CO | |
132.16 | |
132.16 |
14002-80-3 | |
177°C to 178°C | |
Authentic | |
Glass bottle | |
HOCH2C(CH3)2CO2CH3 | |
100g | |
1.04 | |
KJRFTNVYOAGTHK-UHFFFAOYSA-N | |
methyl 3-hydroxy-2,2-dimethylpropanoate | |
84152 | |
98+% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 237-805-3
RUO â Research Use Only