Learn More
Methyl 2,2-dimethyl-3-hydroxypropionate, 98+%
CAS: 14002-80-3 | C6H12O3 | 132.16 g/mol
$147.27 - $147.27
Chemical Identifiers
| CAS | 14002-80-3 |
|---|---|
| Molecular Formula | C6H12O3 |
| Molecular Weight (g/mol) | 132.16 |
| MDL Number | MFCD00009707 |
| InChI Key | KJRFTNVYOAGTHK-UHFFFAOYSA-N |
| Synonym | hydroxypivalic acid methyl ester, methyl 3-hydroxypivalate, methyl hydroxypivalate, methyl 2,2-dimethyl-3-hydroxypropionate, hydroxypivalicacidmethylester, propanoic acid, 3-hydroxy-2,2-dimethyl-, methyl ester, 2,2-dimethyl-3-hydroxypropionic acid methyl ester, 3-hydroxy-2,2-dimethyl-propionic acid methyl ester, methyl 2,2-dimethyl-3-hydroxypropanoate, methyl hydroxylpivalate |
| PubChem CID | 84152 |
| IUPAC Name | methyl 3-hydroxy-2,2-dimethylpropanoate |
| SMILES | COC(=O)C(C)(C)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC210910250
|
Thermo Scientific Chemicals
210910250 |
25 g | Glass bottle |
Each for $147.27
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14002-80-3 | |
| 132.16 | |
| KJRFTNVYOAGTHK-UHFFFAOYSA-N | |
| 84152 | |
| COC(=O)C(C)(C)CO |
| C6H12O3 | |
| MFCD00009707 | |
| hydroxypivalic acid methyl ester, methyl 3-hydroxypivalate, methyl hydroxypivalate, methyl 2,2-dimethyl-3-hydroxypropionate, hydroxypivalicacidmethylester, propanoic acid, 3-hydroxy-2,2-dimethyl-, methyl ester, 2,2-dimethyl-3-hydroxypropionic acid methyl ester, 3-hydroxy-2,2-dimethyl-propionic acid methyl ester, methyl 2,2-dimethyl-3-hydroxypropanoate, methyl hydroxylpivalate | |
| methyl 3-hydroxy-2,2-dimethylpropanoate |
Specifications
| 14002-80-3 | |
| 1.0400g/mL | |
| 76°C | |
| 98% min. (GC) | |
| C6H12O3 | |
| MFCD00009707 | |
| 03, 331 | |
| hydroxypivalic acid methyl ester, methyl 3-hydroxypivalate, methyl hydroxypivalate, methyl 2,2-dimethyl-3-hydroxypropionate, hydroxypivalicacidmethylester, propanoic acid, 3-hydroxy-2,2-dimethyl-, methyl ester, 2,2-dimethyl-3-hydroxypropionic acid methyl ester, 3-hydroxy-2,2-dimethyl-propionic acid methyl ester, methyl 2,2-dimethyl-3-hydroxypropanoate, methyl hydroxylpivalate | |
| COC(=O)C(C)(C)CO | |
| 132.16 | |
| 132.16 | |
| Liquid |
| Colorless | |
| 177°C to 178°C | |
| Authentic | |
| Glass bottle | |
| HOCH2C(CH3)2CO2CH3 | |
| 25 g | |
| 1.04 | |
| KJRFTNVYOAGTHK-UHFFFAOYSA-N | |
| methyl 3-hydroxy-2,2-dimethylpropanoate | |
| 84152 | |
| 98+% | |
| Methyl 2,2-dimethyl-3-hydroxypropionate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 237-805-3
RUO – Research Use Only