missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Amino-5-methylbenzoate 98.0+%, TCI America™
$123.03 - $417.56
Chemical Identifiers
| CAS | 18595-16-9 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.19 |
| MDL Number | MFCD00661547 |
| InChI Key | MDHYFUPTSWXVIA-UHFFFAOYSA-N |
| Synonym | 2-Amino-5-methylbenzoic Acid Methyl Ester, Methyl 6-Amino-m-toluate, 6-Amino-m-toluic Acid Methyl Ester, Methyl 5-Methylanthranilate, 5-Methylanthranilic Acid Methyl Ester |
| PubChem CID | 1201104 |
| IUPAC Name | methyl 2-amino-5-methylbenzoate |
| SMILES | COC(=O)C1=C(N)C=CC(C)=C1 |
Chemical Identifiers
| 18595-16-9 | |
| 165.19 | |
| MDHYFUPTSWXVIA-UHFFFAOYSA-N | |
| 1201104 | |
| COC(=O)C1=C(N)C=CC(C)=C1 |
| C9H11NO2 | |
| MFCD00661547 | |
| 2-Amino-5-methylbenzoic Acid Methyl Ester, Methyl 6-Amino-m-toluate, 6-Amino-m-toluic Acid Methyl Ester, Methyl 5-Methylanthranilate, 5-Methylanthranilic Acid Methyl Ester | |
| methyl 2-amino-5-methylbenzoate |
Specifications
| 18595-16-9 | |
| White-Yellow | |
| MFCD00661547 | |
| 2-Amino-5-methylbenzoic Acid Methyl Ester, Methyl 6-Amino-m-toluate, 6-Amino-m-toluic Acid Methyl Ester, Methyl 5-Methylanthranilate, 5-Methylanthranilic Acid Methyl Ester | |
| COC(=O)C1=C(N)C=CC(C)=C1 | |
| 165.19 | |
| 165.19 | |
| Crystalline Powder |
| 63°C | |
| C9H11NO2 | |
| 5 g | |
| MDHYFUPTSWXVIA-UHFFFAOYSA-N | |
| methyl 2-amino-5-methylbenzoate | |
| 1201104 | |
| ≥98.0% (GC,T) | |
| Methyl 2-Amino-5-methylbenzoate |
Safety and Handling
TSCA : No