missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Aminonicotinate 97.0+%, TCI America™

$50.17 - $134.95

Chemical Identifiers

CAS 14667-47-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00128872
InChI Key NZZDEODTCXHCRS-UHFFFAOYSA-N
PubChem CID 351633
IUPAC Name methyl 2-aminopyridine-3-carboxylate
SMILES COC(=O)C1=CC=CN=C1N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21251G
View Documents
TCI America
M21251G
1 g
Each for $50.17
 
M21255G
View Documents
TCI America
M21255G
5 g
Each for $134.95
 
Description

Description

Specifications

Chemical Identifiers

14667-47-1
152.15
NZZDEODTCXHCRS-UHFFFAOYSA-N
methyl 2-aminopyridine-3-carboxylate
C7H8N2O2
MFCD00128872
351633
COC(=O)C1=CC=CN=C1N

Specifications

14667-47-1
White-Yellow
MFCD00128872
methyl 2-aminonicotinate, 2-amino-nicotinic acid methyl ester, 2-aminonicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl2-aminonicotinate, pubchem16778, methyl?2-aminonicotinate, methyl 2-amino-nicotinate, acmc-1c0pk, aminonicotinsauremethylester
COC(=O)C1=CC=CN=C1N
152.15
152.15
Crystalline Powder
84°C
C7H8N2O2
1 g
NZZDEODTCXHCRS-UHFFFAOYSA-N
methyl 2-aminopyridine-3-carboxylate
351633
≥97.0% (GC,T)
Methyl 2-Aminonicotinate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'