missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-aminopyridine-4-carboxylate, 98%, Thermo Scientific™

Chemical Identifiers

CAS 6937-03-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD04039316
InChI Key SVWWNEYBEFASMP-UHFFFAOYSA-N
Synonym n-2-methoxy-5-methylphenyl benzenesulfonamide, 2'-methoxy-5'-methylbenzenesulfonanilide, benzenesulfonamide, n-2-methoxy-5-methylphenyl, 2-methoxy-5-methylphenyl phenylsulfonyl amine
PubChem CID 248884
IUPAC Name methyl 2-aminopyridine-4-carboxylate
SMILES COC(=O)C1=CC=NC(N)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC430060010
SDS View Documents
Thermo Scientific Chemicals
430060010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC430060050
SDS View Documents
Thermo Scientific Chemicals
430060050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6937-03-7
152.15
SVWWNEYBEFASMP-UHFFFAOYSA-N
248884
COC(=O)C1=CC=NC(N)=C1
C7H8N2O2
MFCD04039316
n-2-methoxy-5-methylphenyl benzenesulfonamide, 2'-methoxy-5'-methylbenzenesulfonanilide, benzenesulfonamide, n-2-methoxy-5-methylphenyl, 2-methoxy-5-methylphenyl phenylsulfonyl amine
methyl 2-aminopyridine-4-carboxylate

Specifications

6937-03-7
97.5% min. (GC)
C7H8N2O2
1g
SVWWNEYBEFASMP-UHFFFAOYSA-N
methyl 2-aminopyridine-4-carboxylate
236984
98%
Authentic
Glass bottle
MFCD04039316
methyl 2-aminoisonicotinate, 2-aminopyridine-4-carboxylic acid methyl ester, 2-aminoisonicotinic acid methyl ester, 4-methoxycarbonyl pyridin-2-amine, 4-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl 2-amino-4-pyridinecarboxylate, methyl-2-aminopyridine-4-carboxylate, 2-amino-4-pyridinecarboxylic acid methylester, 2-amino-4-pyridinecarboxylic acid methyl ester, methyl-2-aminoisonicotinat
COC(=O)C1=CC(=NC=C1)N
152.15
152.15
Methyl 2-aminopyridine-4-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only