missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-bromo-3-methylbenzoate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6183122

 View more versions of this product

Catalog No. AAH6183122


Explore available promotions

Description

Description

Methyl 2-bromo-3-methylbenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromo-3-methylbenzoate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Methyl 2-bromo-3-methylbenzoate
Red
MFCD00144762
4177165
Slightly soluble in water.
COC(=O)C1=C(Br)C(C)=CC=C1
229.07
229.07
Liquid
131001-86-0
C9H9BrO2
100 g
2-bromo-3-methyl-benzoic acid methyl ester, methyl2-bromo-3-methylbenzoate, benzoic acid, 2-bromo-3-methyl-, methyl ester, 2-bromo-3-methylbenzoic acid methyl ester, methyl 2-bromo-3-methyl-benzoate, pubchem3966, acmc-1c3gk, methyl 2-bromo-m-toluate, intermediates-zcf02010, ksc495c9h
QAOFGUXVDAZKBW-UHFFFAOYSA-N
methyl 2-bromo-3-methylbenzoate
10823279
98%

Chemical Identifiers

131001-86-0
229.07
QAOFGUXVDAZKBW-UHFFFAOYSA-N
10823279
COC(=O)C1=C(Br)C(C)=CC=C1
C9H9BrO2
MFCD00144762
2-bromo-3-methyl-benzoic acid methyl ester, methyl2-bromo-3-methylbenzoate, benzoic acid, 2-bromo-3-methyl-, methyl ester, 2-bromo-3-methylbenzoic acid methyl ester, methyl 2-bromo-3-methyl-benzoate, pubchem3966, acmc-1c3gk, methyl 2-bromo-m-toluate, intermediates-zcf02010, ksc495c9h
methyl 2-bromo-3-methylbenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only