missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-chloro-6-methoxypyridine-4-carboxylate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3343814

 View more versions of this product

Catalog No. AAH3343814



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl 2-chloro-6-methoxypyridine-4-carboxylate
103°C to 105°C
MFCD00173929
methyl 2-chloro-6-methoxyisonicotinate, 2-chloro-6-methoxy-isonicotinicacidmethylester, 2-chloro-6-methoxy-4-methoxycarbonyl pyridine, methylchloromethoxyisonicotinate, methyl-2-chloro-6-methoxyisonicotinate, 2-chloro-6-methoxy-isonicotinic acid methyl ester, methyl 2-chloro-6-methoxypyridin-4-yl carboxylate, 2-chloro-6-methoxy-isonicotinic acidmethyl ester, 4-pyridinecarboxylic acid, 2-chloro-6-methoxy-, methyl ester
COC1=NC(=CC(=C1)C(=O)OC)Cl
201.606
201.61
42521-10-8
C8H8ClNO3
25 g
DAPAXVAUEVRBGS-UHFFFAOYSA-N
methyl 2-chloro-6-methoxypyridine-4-carboxylate
13339618
98%

Chemical Identifiers

42521-10-8
201.606
DAPAXVAUEVRBGS-UHFFFAOYSA-N
13339618
COC1=NC(=CC(=C1)C(=O)OC)Cl
C8H8ClNO3
MFCD00173929
methyl 2-chloro-6-methoxyisonicotinate, 2-chloro-6-methoxy-isonicotinicacidmethylester, 2-chloro-6-methoxy-4-methoxycarbonyl pyridine, methylchloromethoxyisonicotinate, methyl-2-chloro-6-methoxyisonicotinate, 2-chloro-6-methoxy-isonicotinic acid methyl ester, methyl 2-chloro-6-methoxypyridin-4-yl carboxylate, 2-chloro-6-methoxy-isonicotinic acidmethyl ester, 4-pyridinecarboxylic acid, 2-chloro-6-methoxy-, methyl ester
methyl 2-chloro-6-methoxypyridine-4-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only