missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Chloropropionate 95.0+%, TCI America™

$59.65 - $264.70

Chemical Identifiers

CAS 17639-93-9
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.55
MDL Number MFCD00000863
InChI Key JLEJCNOTNLZCHQ-UHFFFAOYNA-N
Synonym methyl 2-chloropropionate, propanoic acid, 2-chloro-, methyl ester, 2-chloropropionic acid methyl ester, propionic acid, 2-chloro-, methyl ester, methyl alpha-chloropropionate, methyl .alpha.-chloropropionate, .alpha.-chloropropionic acid methyl ester, propanoic acid, 2-chloro-, methyl ester, s, ch3chclc o och3, acmc-1bl6f
PubChem CID 28659
IUPAC Name methyl 2-chloropropanoate
SMILES COC(=O)C(C)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C097025ML
View Documents
TCI America
C097025ML
25 mL N/A
Only null left
 
C0970100ML
View Documents
TCI America
C0970100ML
100 mL N/A
Only null left
 
C0970500ML
View Documents
TCI America
C0970500ML
500 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

17639-93-9
122.55
JLEJCNOTNLZCHQ-UHFFFAOYNA-N
28659
COC(=O)C(C)Cl
C4H7ClO2
MFCD00000863
methyl 2-chloropropionate, propanoic acid, 2-chloro-, methyl ester, 2-chloropropionic acid methyl ester, propionic acid, 2-chloro-, methyl ester, methyl alpha-chloropropionate, methyl .alpha.-chloropropionate, .alpha.-chloropropionic acid methyl ester, propanoic acid, 2-chloro-, methyl ester, s, ch3chclc o och3, acmc-1bl6f
methyl 2-chloropropanoate

Specifications

17639-93-9
132°C
MFCD00000863
2933
JLEJCNOTNLZCHQ-UHFFFAOYNA-N
methyl 2-chloropropanoate
28659
≥95.0% (GC)
Methyl 2-Chloropropionate
Colorless
C4H7ClO2
25 mL
methyl 2-chloropropionate, propanoic acid, 2-chloro-, methyl ester, 2-chloropropionic acid methyl ester, propionic acid, 2-chloro-, methyl ester, methyl alpha-chloropropionate, methyl .alpha.-chloropropionate, .alpha.-chloropropionic acid methyl ester, propanoic acid, 2-chloro-, methyl ester, s, ch3chclc o och3, acmc-1bl6f
COC(=O)C(C)Cl
122.55
122.55
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3084

RTECSNumber : UE9100000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'