Learn More
Methyl 2-fluoro-4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
$310.45 - $1124.93
Chemical Identifiers
CAS | 197507-22-5 |
---|---|
Molecular Formula | C8H7FO3 |
Molecular Weight (g/mol) | 170.139 |
MDL Number | MFCD09038282 |
InChI Key | WYSPMXSNCAFCFV-UHFFFAOYSA-N |
Synonym | 2-fluoro-4-hydroxy-benzoic acid methyl ester, 2-fluoro-4-hydroxybenzoic acid methyl ester, benzoic acid, 2-fluoro-4-hydroxy-, methyl ester, methyl-2-fluoro4-hydroxybenzoate, methyl2-fluoro-4-hydroxybenzoate, methyl-2-fluoro-4-hydroxybenzoate, methyl 2-fluoro-4-hydroxy-benzoate, 2-fluoro-4-hyroxybenzoic acid methyl ester |
PubChem CID | 22463192 |
IUPAC Name | methyl 2-fluoro-4-hydroxybenzoate |
SMILES | COC(=O)C1=C(C=C(C=C1)O)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6167503
|
Thermo Scientific Chemicals
H6167503 |
1 g |
Each for $310.45
|
|
||||||
AAH6167506
|
Thermo Scientific Chemicals
H6167506 |
5 g |
Each for $1,124.93
|
|
||||||
Description
Methyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 2-fluoro-4-hydroxybenzoate can be used in the preparation of compounds, with the ability to inhibit cytosolic phospholipase A2α. It is also an intermediate in the synthesis of 3-(2-fluoro-4-hydroxyphenyl)-5-(trifluoromethyl) -4-isoxazolecarboxylic acid methyl ester (F591865).
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
197507-22-5 | |
170.139 | |
WYSPMXSNCAFCFV-UHFFFAOYSA-N | |
22463192 | |
COC(=O)C1=C(C=C(C=C1)O)F |
C8H7FO3 | |
MFCD09038282 | |
2-fluoro-4-hydroxy-benzoic acid methyl ester, 2-fluoro-4-hydroxybenzoic acid methyl ester, benzoic acid, 2-fluoro-4-hydroxy-, methyl ester, methyl-2-fluoro4-hydroxybenzoate, methyl2-fluoro-4-hydroxybenzoate, methyl-2-fluoro-4-hydroxybenzoate, methyl 2-fluoro-4-hydroxy-benzoate, 2-fluoro-4-hyroxybenzoic acid methyl ester | |
methyl 2-fluoro-4-hydroxybenzoate |
Specifications
197507-22-5 | |
MFCD09038282 | |
2-fluoro-4-hydroxy-benzoic acid methyl ester, 2-fluoro-4-hydroxybenzoic acid methyl ester, benzoic acid, 2-fluoro-4-hydroxy-, methyl ester, methyl-2-fluoro4-hydroxybenzoate, methyl2-fluoro-4-hydroxybenzoate, methyl-2-fluoro-4-hydroxybenzoate, methyl 2-fluoro-4-hydroxy-benzoate, 2-fluoro-4-hyroxybenzoic acid methyl ester | |
WYSPMXSNCAFCFV-UHFFFAOYSA-N | |
methyl 2-fluoro-4-hydroxybenzoate | |
22463192 | |
98% |
C8H7FO3 | |
1 g | |
Slightly soluble in water. | |
COC(=O)C1=C(C=C(C=C1)O)F | |
170.139 | |
170.14 | |
Methyl 2-fluoro-4-hydroxybenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only