missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxy-3-methylbenzoate, 97%

CAS: 23287-26-5 | C9H10O3 | 166.18 g/mol

$87.74 - $87.74

Chemical Identifiers

CAS 23287-26-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00020051
InChI Key SUHLUMKZPUMAFP-UHFFFAOYSA-N
Synonym methyl 3-methylsalicylate, methyl o-cresotinate, benzoic acid, 2-hydroxy-3-methyl-, methyl ester, levegal pt, 2-hydroxy-3-methylbenzoic acid methyl ester, 2,3-cresotic acid, methyl ester, methyl-3-methylsalicylate, methyl 2-hydroxy-3-methyl-benzoate, methyl o-cresotate, 3-methylsalicylic acid methyl ester
PubChem CID 31835
IUPAC Name methyl 2-hydroxy-3-methylbenzoate
SMILES COC(=O)C1=CC=CC(C)=C1O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2134104
View Documents
Thermo Scientific Chemicals
B2134104
2 g N/A
Only null left
 
Description

Description

Methyl 2-hydroxy-3-methylbenzoate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic intermediate. Some other applications include it is used to produce other chemicals. For example, it is used to produce 2-(2-Hydroxy-3-methyl-phenyl)-propan-2-ol. The reaction occurs with reagent Tetrahydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-3-methylbenzoate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic intermediate. Some other applications include it is used to produce other chemicals. For example, it is used to produce 2-(2-Hydroxy-3-methyl-phenyl)-propan-2-ol. The reaction occurs with reagent Tetrahydrofuran.

Solubility
Soluble in ethanol, fixed Oils, Propylene Glycol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic. Stable under normal temperatures and pressures.
Specifications

Chemical Identifiers

23287-26-5
166.18
SUHLUMKZPUMAFP-UHFFFAOYSA-N
31835
COC(=O)C1=CC=CC(C)=C1O
C9H10O3
MFCD00020051
methyl 3-methylsalicylate, methyl o-cresotinate, benzoic acid, 2-hydroxy-3-methyl-, methyl ester, levegal pt, 2-hydroxy-3-methylbenzoic acid methyl ester, 2,3-cresotic acid, methyl ester, methyl-3-methylsalicylate, methyl 2-hydroxy-3-methyl-benzoate, methyl o-cresotate, 3-methylsalicylic acid methyl ester
methyl 2-hydroxy-3-methylbenzoate

Specifications

23287-26-5
234°C to 236°C
MFCD00020051
methyl 3-methylsalicylate, methyl o-cresotinate, benzoic acid, 2-hydroxy-3-methyl-, methyl ester, levegal pt, 2-hydroxy-3-methylbenzoic acid methyl ester, 2,3-cresotic acid, methyl ester, methyl-3-methylsalicylate, methyl 2-hydroxy-3-methyl-benzoate, methyl o-cresotate, 3-methylsalicylic acid methyl ester
SUHLUMKZPUMAFP-UHFFFAOYSA-N
methyl 2-hydroxy-3-methylbenzoate
31835
97%
28°C to 30°C
C9H10O3
2 g
Soluble in ethanol,fixed Oils,Propylene Glycol. Insoluble in water.
COC(=O)C1=CC=CC(C)=C1O
166.18
166.18
Methyl 2-hydroxy-3-methylbenzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 245-557-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only