missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-(methylthio)benzoate, 98%, Thermo Scientific Chemicals

$86.71 - $337.73

Chemical Identifiers

CAS 3704-28-7
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00068039
InChI Key CPQDZXPLQXZJGF-UHFFFAOYSA-N
PubChem CID 279586
IUPAC Name methyl 2-methylsulfanylbenzoate
SMILES COC(=O)C1=CC=CC=C1SC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1090503
View Documents
Thermo Scientific Chemicals
A1090503
1 g
Each for $86.71
 
AAA1090506
View Documents
Thermo Scientific Chemicals
A1090506
5 g
Each for $337.73
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3704-28-7
182.237
CPQDZXPLQXZJGF-UHFFFAOYSA-N
methyl 2-methylsulfanylbenzoate
C9H10O2S
MFCD00068039
279586
COC(=O)C1=CC=CC=C1SC

Specifications

3704-28-7
Stench
MFCD00068039
2084294
CPQDZXPLQXZJGF-UHFFFAOYSA-N
methyl 2-methylsulfanylbenzoate
279586
98%
64°C to 66°C
C9H10O2S
1 g
methyl 2-methylthio benzoate, methyl 2-methylsulfanyl benzoate, benzoic acid, 2-methylthio-, methyl ester, benzoic acid,2-methylthio-, methyl ester, 2-methylthiobenzoic acid, methyl ester, acmc-1cnec, methyl o-methylthiosalicylate, methyl-2-methylthio benzoate, methyl 2-methylsulfanyl benzoate #, 2-methylthio benzoic acid methyl ester
COC(=O)C1=CC=CC=C1SC
182.237
182.24
Methyl 2- (methylthio)benzoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only