missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3,5-dimethylbenzoate, 98%, Thermo Scientific Chemicals

$80.71 - $241.64

Chemical Identifiers

CAS 25081-39-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00077971
InChI Key PEVXENGLERTHJE-UHFFFAOYSA-N
PubChem CID 32786
IUPAC Name methyl 3,5-dimethylbenzoate
SMILES CC1=CC(=CC(=C1)C(=O)OC)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1228706
View Documents
Thermo Scientific Chemicals
A1228706
5 g
Each for $80.71
 
AAA1228714
View Documents
Thermo Scientific Chemicals
A1228714
25 g
Each for $241.64
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25081-39-4
164.204
PEVXENGLERTHJE-UHFFFAOYSA-N
methyl 3,5-dimethylbenzoate
C10H12O2
MFCD00077971
32786
CC1=CC(=CC(=C1)C(=O)OC)C

Specifications

31°C to 33°C
1.027
5 g
MFCD00077971
PEVXENGLERTHJE-UHFFFAOYSA-N
methyl 3,5-dimethylbenzoate
32786
98%
25081-39-4
239°C to 240°C
C10H12O2
benzoic acid, 3,5-dimethyl-, methyl ester, 3,5-dimethylbenzoic acid methyl ester, acmc-1cmfo, methyl-3,5-dimethylbenzoate, methyl-3,5-dimethyl-benzoate, 3,5-dimethylbenzoic acid methyl, methyl 3,5-dimethylbenzoate, 3,5-ch3 2-c6h3-cooch3, benzoic acid,3,5-dimethyl-, methyl ester, 3,5-dimethyl-benzoic acid methyl ester
CC1=CC(=CC(=C1)C(=O)OC)C
164.204
164.21
Methyl 3,5-dimethylbenzoate
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only