missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Amino-2-fluorobenzoate 96.0+%, TCI America™

$171.29 - $505.19

Chemical Identifiers

CAS 1195768-18-3
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.155
MDL Number MFCD12923265
InChI Key UOYDNSRSUSNCKS-UHFFFAOYSA-N
Synonym 3-Amino-2-fluorobenzoic Acid Methyl Ester
PubChem CID 46311080
IUPAC Name methyl 3-amino-2-fluorobenzoate
SMILES COC(=O)C1=C(C(=CC=C1)N)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M2868200MG
View Documents
TCI America
M2868200MG
200 mg
Each for $171.29
 
M28681G
View Documents
TCI America
M28681G
1 g
Each for $505.19
 
Description

Description

Specifications

Chemical Identifiers

1195768-18-3
169.155
UOYDNSRSUSNCKS-UHFFFAOYSA-N
46311080
COC(=O)C1=C(C(=CC=C1)N)F
C8H8FNO2
MFCD12923265
3-Amino-2-fluorobenzoic Acid Methyl Ester
methyl 3-amino-2-fluorobenzoate

Specifications

1195768-18-3
C8H8FNO2
200 mg
UOYDNSRSUSNCKS-UHFFFAOYSA-N
methyl 3-amino-2-fluorobenzoate
46311080
≥96.0% (GC,T)
Methyl 3-Amino-2-fluorobenzoate
Yellow
MFCD12923265
3-Amino-2-fluorobenzoic Acid Methyl Ester
COC(=O)C1=C(C(=CC=C1)N)F
169.155
169.16
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'