Learn More
Methyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate, 97%, Thermo Scientific Chemicals
$295.84 - $295.84
Chemical Identifiers
CAS | 129332-45-2 |
---|---|
Molecular Formula | C8H8N2O2S2 |
Molecular Weight (g/mol) | 228.284 |
MDL Number | MFCD00052752 |
InChI Key | VBXBCNWDAJLZOC-UHFFFAOYSA-N |
Synonym | methyl 3-amino-4-cyano-5-methylthio thiophene-2-carboxylate, methyl 3-amino-4-cyano-5-methylsulfanyl thiophene-2-carboxylate, methyl3-amino-4-cyano-5-methylthio thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-amino-4-cyano-5-methylthio-, methyl ester, 3-amino-4-cyano-5-methylsulfanylthiophene-2-carboxylic acid methyl ester, methyl 3-azanyl-4-cyano-5-methylsulfanyl-thiophene-2-carboxylate, 3-amino-4-cyano-5-methylthio-2-thiophenecarboxylic acid methyl ester, maybridge1_009000, acmc-1c12n, methyl 3-amino-4-cyano-5-methylthiothiophene-2-carboxylate |
PubChem CID | 2778604 |
IUPAC Name | methyl 3-amino-4-cyano-5-methylsulfanylthiophene-2-carboxylate |
SMILES | COC(=O)C1=C(C(=C(S1)SC)C#N)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH2623309
|
Thermo Scientific Chemicals
H2623309 |
10 g |
Each for $295.84
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
129332-45-2 | |
228.284 | |
VBXBCNWDAJLZOC-UHFFFAOYSA-N | |
2778604 | |
COC(=O)C1=C(C(=C(S1)SC)C#N)N |
C8H8N2O2S2 | |
MFCD00052752 | |
methyl 3-amino-4-cyano-5-methylthio thiophene-2-carboxylate, methyl 3-amino-4-cyano-5-methylsulfanyl thiophene-2-carboxylate, methyl3-amino-4-cyano-5-methylthio thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-amino-4-cyano-5-methylthio-, methyl ester, 3-amino-4-cyano-5-methylsulfanylthiophene-2-carboxylic acid methyl ester, methyl 3-azanyl-4-cyano-5-methylsulfanyl-thiophene-2-carboxylate, 3-amino-4-cyano-5-methylthio-2-thiophenecarboxylic acid methyl ester, maybridge1_009000, acmc-1c12n, methyl 3-amino-4-cyano-5-methylthiothiophene-2-carboxylate | |
methyl 3-amino-4-cyano-5-methylsulfanylthiophene-2-carboxylate |
Specifications
129332-45-2 | |
C8H8N2O2S2 | |
10 g | |
methyl 3-amino-4-cyano-5-methylthio thiophene-2-carboxylate, methyl 3-amino-4-cyano-5-methylsulfanyl thiophene-2-carboxylate, methyl3-amino-4-cyano-5-methylthio thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-amino-4-cyano-5-methylthio-, methyl ester, 3-amino-4-cyano-5-methylsulfanylthiophene-2-carboxylic acid methyl ester, methyl 3-azanyl-4-cyano-5-methylsulfanyl-thiophene-2-carboxylate, 3-amino-4-cyano-5-methylthio-2-thiophenecarboxylic acid methyl ester, maybridge1_009000, acmc-1c12n, methyl 3-amino-4-cyano-5-methylthiothiophene-2-carboxylate | |
COC(=O)C1=C(C(=C(S1)SC)C#N)N | |
228.284 | |
228.28 | |
Methyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate |
205°C to 208°C | |
MFCD00052752 | |
UN3439 | |
VBXBCNWDAJLZOC-UHFFFAOYSA-N | |
methyl 3-amino-4-cyano-5-methylsulfanylthiophene-2-carboxylate | |
2778604 | |
97% |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only