Learn More
Methyl 3-amino-4-iodobenzoate, 97%
CAS: 412947-54-7 | C8H8INO2 | 277.061 g/mol
$102.55 - $377.78
Chemical Identifiers
| CAS | 412947-54-7 |
|---|---|
| Molecular Formula | C8H8INO2 |
| Molecular Weight (g/mol) | 277.061 |
| MDL Number | MFCD08064225 |
| InChI Key | WJEBNIVVLJEIKE-UHFFFAOYSA-N |
| Synonym | 3-amino-4-iodobenzoic acid methyl ester, benzoic acid, 3-amino-4-iodo-, methyl ester, methyl3-amino-4-iodobenzoate, pubchem10509, acmc-209ygt, ksc495i4l, methyl-3-amino-4-iodobenzoate, methyl 3-amino-4-iodo-benzoate, 3-amino-4-iodo-benzoic acid methyl ester |
| PubChem CID | 21628210 |
| IUPAC Name | methyl 3-amino-4-iodobenzoate |
| SMILES | COC(=O)C1=CC(=C(C=C1)I)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2569903
|
Thermo Scientific Chemicals
B2569903 |
1 g |
Each for $102.55
|
|
|||||
|
AAB2569906
|
Thermo Scientific Chemicals
B2569906 |
5 g |
Each for $377.78
|
|
|||||
Description
Methyl 3-amino-4-iodobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3-amino-4-iodobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.
Chemical Identifiers
| 412947-54-7 | |
| 277.061 | |
| WJEBNIVVLJEIKE-UHFFFAOYSA-N | |
| 21628210 | |
| COC(=O)C1=CC(=C(C=C1)I)N |
| C8H8INO2 | |
| MFCD08064225 | |
| 3-amino-4-iodobenzoic acid methyl ester, benzoic acid, 3-amino-4-iodo-, methyl ester, methyl3-amino-4-iodobenzoate, pubchem10509, acmc-209ygt, ksc495i4l, methyl-3-amino-4-iodobenzoate, methyl 3-amino-4-iodo-benzoate, 3-amino-4-iodo-benzoic acid methyl ester | |
| methyl 3-amino-4-iodobenzoate |
Specifications
| 412947-54-7 | |
| C8H8INO2 | |
| 1 g | |
| 3-amino-4-iodobenzoic acid methyl ester, benzoic acid, 3-amino-4-iodo-, methyl ester, methyl3-amino-4-iodobenzoate, pubchem10509, acmc-209ygt, ksc495i4l, methyl-3-amino-4-iodobenzoate, methyl 3-amino-4-iodo-benzoate, 3-amino-4-iodo-benzoic acid methyl ester | |
| WJEBNIVVLJEIKE-UHFFFAOYSA-N | |
| methyl 3-amino-4-iodobenzoate | |
| 21628210 | |
| 97% |
| 134°C to 136°C | |
| MFCD08064225 | |
| Light sensitive | |
| Slightly soluble in water. | |
| COC(=O)C1=CC(=C(C=C1)I)N | |
| 277.061 | |
| 277.06 | |
| Methyl 3-amino-4-iodobenzoate |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only