missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-Bromo-2-(bromomethyl)propionate 97.0+%, TCI America™

$167.87 - $635.95

Chemical Identifiers

CAS 22262-60-8
Molecular Formula C5H8Br2O2
Molecular Weight (g/mol) 259.925
MDL Number MFCD00011534
InChI Key USXVPPOARMSYGY-UHFFFAOYSA-N
PubChem CID 140895
IUPAC Name methyl 3-bromo-2-(bromomethyl)propanoate
SMILES COC(=O)C(CBr)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20665G
View Documents
TCI America
M20665G
5 g
Each for $167.87
 
M206625G
View Documents
TCI America
M206625G
25 g
Each for $635.95
 
Description

Description

Specifications

Chemical Identifiers

22262-60-8
259.925
USXVPPOARMSYGY-UHFFFAOYSA-N
methyl 3-bromo-2-(bromomethyl)propanoate
C5H8Br2O2
MFCD00011534
140895
COC(=O)C(CBr)CBr

Specifications

22262-60-8
C5H8Br2O2
5 g
USXVPPOARMSYGY-UHFFFAOYSA-N
methyl 3-bromo-2-(bromomethyl)propanoate
140895
≥97.0% (GC)
Methyl 3-Bromo-2-(bromomethyl)propionate
62°C
MFCD00011534
methyl 3-bromo-2-bromomethyl propionate, methyl 3-bromo-2-bromomethyl propanoate, methyl3-bromo-2-bromomethyl propionate, methyl bis bromomethyl acetate, methyl 2-bis-bromomethyl-acetate, propanoic acid, 3-bromo-2-bromomethyl-, methyl ester, propanoic acid,3-bromo-2-bromomethyl-, methyl ester, zlchem 71, acmc-1chw2, 3-bromo-2-bromomethyl-propionic acid methyl ester
COC(=O)C(CBr)CBr
259.925
259.93
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'