missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-bromopropionate, 97%, Thermo Scientific Chemicals

$124.01 - $325.05

Chemical Identifiers

CAS 3395-91-3
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00000250
InChI Key KQEVIFKPZOGBMZ-UHFFFAOYSA-N
Synonym methyl 3-bromopropionate, propanoic acid, 3-bromo-, methyl ester, methyl beta-bromopropionate, 3-bromopropionic acid methyl ester, propionic acid, 3-bromo-, methyl ester, 3-bromopropanoic acid methyl ester, unii-y803ch1p4w, beta-bromopropionic acid, methyl ester, methyl .beta.-bromopropionate, 3-bromo-propionic acid methyl ester
PubChem CID 76934
IUPAC Name methyl 3-bromopropanoate
SMILES COC(=O)CCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1412714
View Documents
Thermo Scientific Chemicals
L1412714
25 g
Each for $124.01
 
AAL1412722
View Documents
Thermo Scientific Chemicals
L1412722
100 g
Each for $325.05
 
Description

Description

Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones. It can react with 2-Methyl-1H-benzoimidazole to get 1-(2-Methoxycarbonylethyl)-2-methylbenzimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones. It can react with 2-Methyl-1H-benzoimidazole to get 1-(2-Methoxycarbonylethyl)-2-methylbenzimidazole.

Solubility
Immiscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications

Chemical Identifiers

3395-91-3
167.00
KQEVIFKPZOGBMZ-UHFFFAOYSA-N
76934
COC(=O)CCBr
C4H7BrO2
MFCD00000250
methyl 3-bromopropionate, propanoic acid, 3-bromo-, methyl ester, methyl beta-bromopropionate, 3-bromopropionic acid methyl ester, propionic acid, 3-bromo-, methyl ester, 3-bromopropanoic acid methyl ester, unii-y803ch1p4w, beta-bromopropionic acid, methyl ester, methyl .beta.-bromopropionate, 3-bromo-propionic acid methyl ester
methyl 3-bromopropanoate

Specifications

3395-91-3
58°C to 60°C (12 mmHg)
C4H7BrO2
MFCD00000250
1742949
Immiscible in water.
COC(=O)CCBr
167.00
167.01
Methyl 3-bromopropionate
1.53
75°C (167°F)
1.458
25 g
methyl 3-bromopropionate, propanoic acid, 3-bromo-, methyl ester, methyl beta-bromopropionate, 3-bromopropionic acid methyl ester, propionic acid, 3-bromo-, methyl ester, 3-bromopropanoic acid methyl ester, unii-y803ch1p4w, beta-bromopropionic acid, methyl ester, methyl .beta.-bromopropionate, 3-bromo-propionic acid methyl ester
KQEVIFKPZOGBMZ-UHFFFAOYSA-N
methyl 3-bromopropanoate
76934
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319-H335

EINECSNumber : 222-247-5

RTECSNumber : UA2451790

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only