missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3-(dimethylamino)-2-isocyanoacrylate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 372020010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 3-(dimethylamino)-2-isocyanoacrylate | |
61.0°C to 67.0°C | |
Authentic | |
Glass bottle | |
1 g | |
Solubility in water: insoluble | |
CN(C)C=C(C(=O)OC)[N+]#[C-] | |
154.17 | |
154.17 | |
Crystalline Powder |
113212-14-9 | |
Yellow | |
95% min. (GC) | |
C7H10N2O2 | |
3-dimethylamino-2-isocyanoacrylic acid methyl ester, methyl e-3-dimethylamino-2-isocyanoprop-2-enoate, methyl 2e-3-dimethylamino-2-isocyanoprop-2-enoate | |
FREGRQYTSPAJDX-AATRIKPKSA-N | |
methyl (E)-3-(dimethylamino)-2-isocyanoprop-2-enoate | |
6387542 | |
96% |
Chemical Identifiers
113212-14-9 | |
154.17 | |
3-dimethylamino-2-isocyanoacrylic acid methyl ester, methyl e-3-dimethylamino-2-isocyanoprop-2-enoate, methyl 2e-3-dimethylamino-2-isocyanoprop-2-enoate | |
methyl (E)-3-(dimethylamino)-2-isocyanoprop-2-enoate |
C7H10N2O2 | |
FREGRQYTSPAJDX-AATRIKPKSA-N | |
6387542 | |
CN(C)C=C(C(=O)OC)[N+]#[C-] |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only