missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-formylbenzoate, 98%

CAS: 52178-50-4 | C9H8O3 | 164.2 g/mol

$391.44 - $391.44

Chemical Identifiers

CAS 52178-50-4
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.2
MDL Number MFCD00189379
InChI Key UVSBCUAQEZINCQ-UHFFFAOYSA-N
Synonym 3-carbomethoxybenzaldehyde, methyl m-formylbenzoate, benzoic acid, 3-formyl-, methyl ester, 3-formylbenzoic acid methyl ester, methyl-3-formylbenzoate, pubchem12631, m-carbomethoxybenzaldehyde, methyl 3-formyl-benzoate, methyl 3-methanoylbenzoate, acmc-1am8j
PubChem CID 639145
IUPAC Name methyl 3-formylbenzoate
SMILES COC(=O)C1=CC=CC(=C1)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC368050050
SDS View Documents
Thermo Scientific Chemicals
368050050
5 g Glass bottle
Each for $391.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

52178-50-4
164.2
UVSBCUAQEZINCQ-UHFFFAOYSA-N
639145
COC(=O)C1=CC=CC(=C1)C=O
C9H8O3
MFCD00189379
3-carbomethoxybenzaldehyde, methyl m-formylbenzoate, benzoic acid, 3-formyl-, methyl ester, 3-formylbenzoic acid methyl ester, methyl-3-formylbenzoate, pubchem12631, m-carbomethoxybenzaldehyde, methyl 3-formyl-benzoate, methyl 3-methanoylbenzoate, acmc-1am8j
methyl 3-formylbenzoate

Specifications

52178-50-4
97.5% min. (GC)
C9H8O3
5 g
Solubility in water: insoluble. Other solubilities: soluble in ethanol
COC(=O)C1=CC=CC(=C1)C=O
164.2
164.2
Methyl 3-formylbenzoate, 98%
48.0°C to 55.0°C
Glass bottle
MFCD00189379
3-carbomethoxybenzaldehyde, methyl m-formylbenzoate, benzoic acid, 3-formyl-, methyl ester, 3-formylbenzoic acid methyl ester, methyl-3-formylbenzoate, pubchem12631, m-carbomethoxybenzaldehyde, methyl 3-formyl-benzoate, methyl 3-methanoylbenzoate, acmc-1am8j
UVSBCUAQEZINCQ-UHFFFAOYSA-N
methyl 3-formylbenzoate
639145
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only