missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-methoxy-4-nitrobenzoate, 98%, Thermo Scientific Chemicals

$108.59 - $400.03

Chemical Identifiers

CAS 5081-37-8
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.173
MDL Number MFCD00051967
InChI Key PVVFEPFLFHDHHK-UHFFFAOYSA-N
Synonym 3-methoxy-4-nitrobenzoic acid methyl ester, benzoic acid, 3-methoxy-4-nitro-, methyl ester, benzoic acid, 5-methoxy-4-nitro-, methyl ester, methyl3-methoxy-4-nitrobenzoate, pubchem4216, acmc-209kq5, ksc275c3p, methylmethoxynitrobenzenecarboxylate, 4-nitro-3-methoxy benzoic acid, 3-methoxy-4-nitro-benzoic acid methyl ester
PubChem CID 287308
IUPAC Name methyl 3-methoxy-4-nitrobenzoate
SMILES COC1=C(C=CC(=C1)C(=O)OC)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2319603
View Documents
Thermo Scientific Chemicals
B2319603
1 g
Each for $108.59
 
AAB2319606
View Documents
Thermo Scientific Chemicals
B2319606
5 g
Each for $400.03
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5081-37-8
211.173
PVVFEPFLFHDHHK-UHFFFAOYSA-N
287308
COC1=C(C=CC(=C1)C(=O)OC)[N+](=O)[O-]
C9H9NO5
MFCD00051967
3-methoxy-4-nitrobenzoic acid methyl ester, benzoic acid, 3-methoxy-4-nitro-, methyl ester, benzoic acid, 5-methoxy-4-nitro-, methyl ester, methyl3-methoxy-4-nitrobenzoate, pubchem4216, acmc-209kq5, ksc275c3p, methylmethoxynitrobenzenecarboxylate, 4-nitro-3-methoxy benzoic acid, 3-methoxy-4-nitro-benzoic acid methyl ester
methyl 3-methoxy-4-nitrobenzoate

Specifications

5081-37-8
C9H9NO5
1 g
3-methoxy-4-nitrobenzoic acid methyl ester, benzoic acid, 3-methoxy-4-nitro-, methyl ester, benzoic acid, 5-methoxy-4-nitro-, methyl ester, methyl3-methoxy-4-nitrobenzoate, pubchem4216, acmc-209kq5, ksc275c3p, methylmethoxynitrobenzenecarboxylate, 4-nitro-3-methoxy benzoic acid, 3-methoxy-4-nitro-benzoic acid methyl ester
COC1=C(C=CC(=C1)C(=O)OC)[N+](=O)[O-]
211.173
211.18
Methyl 3-methoxy-4-nitrobenzoate
83°C to 87°C
MFCD00051967
2856207
PVVFEPFLFHDHHK-UHFFFAOYSA-N
methyl 3-methoxy-4-nitrobenzoate
287308
98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only