Methyl 3-Methyl-4-nitrobenzoate 98.0+%, TCI America™
Supplier:  TCI America M214525G
 View more versions of this product
Catalog No. M214525G
Explore available promotions
Description
Description
Specifications
Specifications
Methyl 3-Methyl-4-nitrobenzoate
81°C
C9H9NO4
25 g
IEFONJKJLZFGKQ-UHFFFAOYSA-N
methyl 3-methyl-4-nitrobenzoate
260927
≥98.0% (GC)
24078-21-5
Yellow
MFCD00085640
benzoic acid, 3-methyl-4-nitro-, methyl ester, 3-methyl-4-nitrobenzoic acid methyl ester, m-toluic acid, 4-nitro-, methyl ester, methyl 3-methyl-4-nitrobenzenecarboxylate, benzoic acid, 3-methyl-4-nitro, methyl ester, methyl3-methyl-4-nitrobenzoate, pubchem2350, acmc-1cfdu, methyl 4-nitro-m-toluate, ksc205m3d
CC1=C(C=CC(=C1)C(=O)OC)[N+](=O)[O-]
195.174
195.17
Crystalline Powder
Chemical Identifiers
24078-21-5
195.174
IEFONJKJLZFGKQ-UHFFFAOYSA-N
260927
CC1=C(C=CC(=C1)C(=O)OC)[N+](=O)[O-]
C9H9NO4
MFCD00085640
benzoic acid, 3-methyl-4-nitro-, methyl ester, 3-methyl-4-nitrobenzoic acid methyl ester, m-toluic acid, 4-nitro-, methyl ester, methyl 3-methyl-4-nitrobenzenecarboxylate, benzoic acid, 3-methyl-4-nitro, methyl ester, methyl3-methyl-4-nitrobenzoate, pubchem2350, acmc-1cfdu, methyl 4-nitro-m-toluate, ksc205m3d
methyl 3-methyl-4-nitrobenzoate
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
TSCA : No