missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3-nitrobenzoate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 168440250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl 3-nitrobenzoate | |
279°C | |
97.5% min. (GC) | |
C8H7NO4 | |
MFCD00007250 | |
09, 378 | |
3-nitrobenzoic acid methyl ester, methyl m-nitrobenzoate, methyl3-nitrobenzoate, benzoic acid, 3-nitro-, methyl ester, m-nitrobenzoic acid, methyl ester, benzoic acid, m-nitro-, methyl ester, 3-nitro-benzoic acid methyl ester, 3-nitro-benzoicacimethylester, acmc-209mxc, methyl 3-nitro-benzoate | |
AXLYJLKKPUICKV-UHFFFAOYSA-N | |
methyl 3-nitrobenzoate | |
69260 | |
98% |
618-95-1 | |
Authentic | |
Plastic bottle | |
O2NC6H4CO2CH3 | |
25g | |
10, 6435 | |
Solubility in water: insoluble | |
COC(=O)C1=CC(=CC=C1)[N+](=O)[O-] | |
181.15 | |
181.15 |
Safety and Handling
EINECSNumber : 210-573-
RUO â Research Use Only