missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-Oxohexanoate 96.0+%, TCI America™
$52.19 - $426.87
Chemical Identifiers
| CAS | 30414-54-1 |
|---|---|
| Molecular Formula | C7H12O3 |
| Molecular Weight (g/mol) | 144.17 |
| MDL Number | MFCD00040995 |
| InChI Key | SJPCQNABHNCLPB-UHFFFAOYSA-N |
| Synonym | methyl butyrylacetate, methyl 3-oxocaproate, 3-oxohexanoic acid methyl ester, hexanoic acid, 3-oxo-, methyl ester, methyl 3-ketohexanoate, butyrylacetic acid methyl ester, 3-oxo-hexanoic acid methyl ester, bem, acmc-1ahmy, ksc497e5t |
| PubChem CID | 121700 |
| IUPAC Name | methyl 3-oxohexanoate |
| SMILES | CCCC(=O)CC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
K003725ML
|
TCI America
K003725ML |
25 mL |
Each for $52.19
|
|
|||||
|
K0037500ML
|
TCI America
K0037500ML |
500 mL |
Each for $426.87
|
|
|||||
Chemical Identifiers
| 30414-54-1 | |
| 144.17 | |
| SJPCQNABHNCLPB-UHFFFAOYSA-N | |
| 121700 | |
| CCCC(=O)CC(=O)OC |
| C7H12O3 | |
| MFCD00040995 | |
| methyl butyrylacetate, methyl 3-oxocaproate, 3-oxohexanoic acid methyl ester, hexanoic acid, 3-oxo-, methyl ester, methyl 3-ketohexanoate, butyrylacetic acid methyl ester, 3-oxo-hexanoic acid methyl ester, bem, acmc-1ahmy, ksc497e5t | |
| methyl 3-oxohexanoate |
Specifications
| 30414-54-1 | |
| 85°C | |
| MFCD00040995 | |
| methyl butyrylacetate, methyl 3-oxocaproate, 3-oxohexanoic acid methyl ester, hexanoic acid, 3-oxo-, methyl ester, methyl 3-ketohexanoate, butyrylacetic acid methyl ester, 3-oxo-hexanoic acid methyl ester, bem, acmc-1ahmy, ksc497e5t | |
| CCCC(=O)CC(=O)OC | |
| 144.17 | |
| 144.17 | |
| Liquid |
| Colorless | |
| C7H12O3 | |
| 25 mL | |
| SJPCQNABHNCLPB-UHFFFAOYSA-N | |
| methyl 3-oxohexanoate | |
| 121700 | |
| ≥96.0% (GC) | |
| Methyl 3-Oxohexanoate |
Safety and Handling
EINECSNumber : (2)-1505
TSCA : No