missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 4-(1,1-Dioxothiomorpholino)benzoate 98.0+%, TCI America™
Supplier: TCI America M15871G
Specifications
Methyl 4-(1,1-Dioxothiomorpholino)benzoate | |
Yellow | |
1 g | |
IMLAPXMJROBNSS-UHFFFAOYSA-N | |
methyl 4-(1,1-dioxo-1,4-thiazinan-4-yl)benzoate | |
2763956 | |
≥98.0% (GC) |
451485-76-0 | |
C12H15NO4S | |
4-(1,1-Dioxothiomorpholino)benzoic Acid Methyl Ester, 4-[4-(Methoxycarbonyl)phenyl]thiomorpholine 1,1-Dioxide | |
COC(=O)C1=CC=C(C=C1)N2CCS(=O)(=O)CC2 | |
269.315 | |
269.32 | |
Crystalline Powder |
Chemical Identifiers
451485-76-0 | |
269.315 | |
4-(1,1-Dioxothiomorpholino)benzoic Acid Methyl Ester, 4-[4-(Methoxycarbonyl)phenyl]thiomorpholine 1,1-Dioxide | |
methyl 4-(1,1-dioxo-1,4-thiazinan-4-yl)benzoate |
C12H15NO4S | |
IMLAPXMJROBNSS-UHFFFAOYSA-N | |
2763956 | |
COC(=O)C1=CC=C(C=C1)N2CCS(=O)(=O)CC2 |
Safety and Handling
TSCA : No