missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,5-dimethoxy-2-nitrobenzoate, 97%

CAS: 26791-93-5 | C10H11NO6 | 241.199 g/mol

$87.90 - $494.29

Chemical Identifiers

CAS 26791-93-5
Molecular Formula C10H11NO6
Molecular Weight (g/mol) 241.199
MDL Number MFCD00007239
InChI Key SYYKLKHBZGFKOC-UHFFFAOYSA-N
Synonym methyl 6-nitroveratrate, benzoic acid, 4,5-dimethoxy-2-nitro-, methyl ester, 6-nitroveratric acid methyl ester, 4,5-dimethoxy-2-nitrobenzoic acid, methyl ester, methyl 4,5-dimethoxy-2-nitro-benzoate, acmc-1cptq, ksc496m9r, methyl 3,4-dimethoxy-6-nitrobenzoate, methyl 3,4-dimethoxy-6-nitro-benzoate, methyl 4,5-dimethoxy-2-nitrobenzoate
PubChem CID 117867
IUPAC Name methyl 4,5-dimethoxy-2-nitrobenzoate
SMILES COC1=C(C=C(C(=C1)C(=O)OC)[N+](=O)[O-])OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1283618
View Documents
Thermo Scientific Chemicals
A1283618
50 g N/A
Only null left
 
AAA1283630
View Documents
Thermo Scientific Chemicals
A1283630
250 g N/A
Only null left
 
AAA1283636
View Documents
Thermo Scientific Chemicals
A1283636
500 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

26791-93-5
241.199
SYYKLKHBZGFKOC-UHFFFAOYSA-N
117867
COC1=C(C=C(C(=C1)C(=O)OC)[N+](=O)[O-])OC
C10H11NO6
MFCD00007239
methyl 6-nitroveratrate, benzoic acid, 4,5-dimethoxy-2-nitro-, methyl ester, 6-nitroveratric acid methyl ester, 4,5-dimethoxy-2-nitrobenzoic acid, methyl ester, methyl 4,5-dimethoxy-2-nitro-benzoate, acmc-1cptq, ksc496m9r, methyl 3,4-dimethoxy-6-nitrobenzoate, methyl 3,4-dimethoxy-6-nitro-benzoate, methyl 4,5-dimethoxy-2-nitrobenzoate
methyl 4,5-dimethoxy-2-nitrobenzoate

Specifications

26791-93-5
C10H11NO6
50 g
methyl 6-nitroveratrate, benzoic acid, 4,5-dimethoxy-2-nitro-, methyl ester, 6-nitroveratric acid methyl ester, 4,5-dimethoxy-2-nitrobenzoic acid, methyl ester, methyl 4,5-dimethoxy-2-nitro-benzoate, acmc-1cptq, ksc496m9r, methyl 3,4-dimethoxy-6-nitrobenzoate, methyl 3,4-dimethoxy-6-nitro-benzoate, methyl 4,5-dimethoxy-2-nitrobenzoate
COC1=C(C=C(C(=C1)C(=O)OC)[N+](=O)[O-])OC
241.199
241.2
Methyl 4,5-dimethoxy-2-nitrobenzoate
142°C to 144°C
MFCD00007239
2381747
SYYKLKHBZGFKOC-UHFFFAOYSA-N
methyl 4,5-dimethoxy-2-nitrobenzoate
117867
97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 248-008-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only