Learn More
Methyl 4,6-dichloronicotinate, 95%
CAS: 65973-52-6 | C7H5Cl2NO2 | 206.02 g/mol
$163.82 - $725.53
Chemical Identifiers
| CAS | 65973-52-6 |
|---|---|
| Molecular Formula | C7H5Cl2NO2 |
| Molecular Weight (g/mol) | 206.02 |
| MDL Number | MFCD04125732 |
| InChI Key | JEJMDUMJSZTJTI-UHFFFAOYSA-N |
| PubChem CID | 2764083 |
| IUPAC Name | methyl 4,6-dichloropyridine-3-carboxylate |
| SMILES | COC(=O)C1=CN=C(Cl)C=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6407706
|
Thermo Scientific Chemicals
H6407706 |
5 g |
Each for $163.82
|
|
|||||
|
AAH6407714
|
Thermo Scientific Chemicals
H6407714 |
25 g |
Each for $725.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 65973-52-6 | |
| 206.02 | |
| JEJMDUMJSZTJTI-UHFFFAOYSA-N | |
| methyl 4,6-dichloropyridine-3-carboxylate |
| C7H5Cl2NO2 | |
| MFCD04125732 | |
| 2764083 | |
| COC(=O)C1=CN=C(Cl)C=C1Cl |
Specifications
| 65973-52-6 | |
| MFCD04125732 | |
| 13279782 | |
| JEJMDUMJSZTJTI-UHFFFAOYSA-N | |
| methyl 4,6-dichloropyridine-3-carboxylate | |
| 2764083 | |
| 95% | |
| Methyl 4,6-dichloronicotinate |
| C7H5Cl2NO2 | |
| 5 g | |
| methyl 4,6-dichloronicotinate, methyl4,6-dichloronicotinate, 4,6-dichloro-nicotinic acid methyl, 4,6-dichloro-nicotinic acid methyl ester, 4,6-dichloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 4,6-dichloro-, methyl ester, 2-p-tolylimidazo 1,2-a pyridine, pubchem12979, ksc494c6d, methyl-4,6-dichloronicotinate | |
| COC(=O)C1=CN=C(Cl)C=C1Cl | |
| 206.02 | |
| 206.02 | |
| Solid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only