Learn More
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, 97%
CAS: 3162-96-7 | C14H18O6 | 282.292 g/mol
$75.38 - $736.14
Chemical Identifiers
| CAS | 3162-96-7 |
|---|---|
| Molecular Formula | C14H18O6 |
| Molecular Weight (g/mol) | 282.292 |
| MDL Number | MFCD00006819 |
| InChI Key | VVSWDMJYIDBTMV-BTZLDLHRSA-N |
| Synonym | 4ar,6s,7r,8r,8as-6-methoxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxine-7,8-diol, methyl 4,6-o-benzylidene-alpha-d-glucopyranoside, methyl 4,6-o-benzylidene-alpha-d-glucopyranoside, 4ar,6s,7r,8r,8as-6-methoxy-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxine-7,8-diol, methyl 4,6-o-benzylidenehexopyranoside #, methyl 4,6-o-benzylidenehexopyranoside #, zlchem 511, a-d-glucopyranoside, a-d-glucopyranoside, glucopyranoside, methyl 4,6-o-benzylidene-, .alpha.-d, glucopyranoside, methyl 4,6-o-benzylidene-, .alpha.-d |
| PubChem CID | 11822086 |
| IUPAC Name | (4aR,6S,7R,8R,8aS)-6-methoxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-7,8-diol |
| SMILES | COC1C(C(C2C(O1)COC(O2)C3=CC=CC=C3)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1341806
|
Thermo Scientific Chemicals
A1341806 |
5 g |
Each for $75.38
|
|
|||||
|
AAA1341814
|
Thermo Scientific Chemicals
A1341814 |
25 g |
Each for $275.38
|
|
|||||
|
AAA1341822
|
Thermo Scientific Chemicals
A1341822 |
100 g |
Each for $736.14
|
|
|||||
Description
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.
Solubility
It is soluble in water, acetone, benzene, ethyl ether
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Chemical Identifiers
| 3162-96-7 | |
| 282.292 | |
| VVSWDMJYIDBTMV-BTZLDLHRSA-N | |
| 11822086 | |
| COC1C(C(C2C(O1)COC(O2)C3=CC=CC=C3)O)O |
| C14H18O6 | |
| MFCD00006819 | |
| 4ar,6s,7r,8r,8as-6-methoxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxine-7,8-diol, methyl 4,6-o-benzylidene-alpha-d-glucopyranoside, methyl 4,6-o-benzylidene-alpha-d-glucopyranoside, 4ar,6s,7r,8r,8as-6-methoxy-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxine-7,8-diol, methyl 4,6-o-benzylidenehexopyranoside #, methyl 4,6-o-benzylidenehexopyranoside #, zlchem 511, a-d-glucopyranoside, a-d-glucopyranoside, glucopyranoside, methyl 4,6-o-benzylidene-, .alpha.-d, glucopyranoside, methyl 4,6-o-benzylidene-, .alpha.-d | |
| (4aR,6S,7R,8R,8aS)-6-methoxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-7,8-diol |
Specifications
| 3162-96-7 | |
| C14H18O6 | |
| 5 g | |
| Hygroscopic | |
| It is soluble in water,acetone,benzene,ethyl ether | |
| +110° (c=2 in Chloroform) | |
| (4aR,6S,7R,8R,8aS)-6-methoxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-7,8-diol | |
| 11822086 | |
| 97% |
| 163°C to 166°C | |
| MFCD00006819 | |
| 1291458 | |
| 4ar,6s,7r,8r,8as-6-methoxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxine-7,8-diol, .alpha.-d-glucopyranoside, methyl 4,6-o-phenylmethylene, methyl 4,6-o-benzylidene-alpha-d-glucopyranoside, 4ar,6s,7r,8r,8as-6-methoxy-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxine-7,8-diol, 4ar,6s,7r,8r,8as-6-methoxy-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxine-7,8-diol, methyl 4,6-o-benzylidenehexopyranoside #, methyl 4,6-o-benzylidene-, a-d-glucopyranoside, methyl 4,6-o-phenylmethylene-, methyl 4,6-o-phenylmethylene-, glucopyranoside, methyl 4,6-o-benzylidene-, .alpha.-d | |
| VVSWDMJYIDBTMV-BTZLDLHRSA-N | |
| COC1C(C(C2C(O1)COC(O2)C3=CC=CC=C3)O)O | |
| 282.292 | |
| 282.29 | |
| Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside |
Safety and Handling
EINECSNumber : 221-615-2
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only