missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-amino-3-nitrobenzoate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3434803

 View more versions of this product

Catalog No. AAH3434803


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl 4-amino-3-nitrobenzoate
188°C to 191°C
MFCD00017562
methyl 4-amino-3-nitrobenzenecarboxylate, benzoic acid, 4-amino-3-nitro-, methyl ester, 4-amino-3-nitro-benzoic acid methyl ester, 4-methoxycarbonyl-2-nitroaniline, 4-amino-3-nitromethylbenzoate, methyl 3-nitro-4-aminobenzoate, 2-amino-5-methoxycarbonyl nitrobenzene, 3-nitro-4-aminobenzoic acid methyl ester, 4-amino-3-nitrobenzoicacidmethylester, acmc-209j8j
COC(=O)C1=CC(=C(C=C1)N)[N+](=O)[O-]
196.162
196.16
3987-92-6
C8H8N2O4
1 g
HNTLUEZVPLRQEV-UHFFFAOYSA-N
methyl 4-amino-3-nitrobenzoate
3941008
97%

Chemical Identifiers

3987-92-6
196.162
HNTLUEZVPLRQEV-UHFFFAOYSA-N
3941008
COC(=O)C1=CC(=C(C=C1)N)[N+](=O)[O-]
C8H8N2O4
MFCD00017562
methyl 4-amino-3-nitrobenzenecarboxylate, benzoic acid, 4-amino-3-nitro-, methyl ester, 4-amino-3-nitro-benzoic acid methyl ester, 4-methoxycarbonyl-2-nitroaniline, 4-amino-3-nitromethylbenzoate, methyl 3-nitro-4-aminobenzoate, 2-amino-5-methoxycarbonyl nitrobenzene, 3-nitro-4-aminobenzoic acid methyl ester, 4-amino-3-nitrobenzoicacidmethylester, acmc-209j8j
methyl 4-amino-3-nitrobenzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only