missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-aminobutyrate hydrochloride, 99%, Thermo Scientific Chemicals

$821.99 - $821.99

Chemical Identifiers

CAS 13031-60-2
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00043270
InChI Key WPGPRLVPWACBHW-UHFFFAOYSA-N
Synonym methyl 4-aminobutanoate hydrochloride, methyl 4-aminobutyrate hydrochloride, methyl 4-aminobutanoate hcl, methyl 4-aminobutyrate hcl, h-gamma-abu-ome.hcl, 4-aminobutyric acid methyl ester hydrochloride, 4-aminobutyric acid methyl ester, hydrochloride, 4-amino-n-butyric acid methyl ester hydrochloride, butanoic acid, 4-amino-, methyl ester, hydrochloride, 4-amino-butyric acid methyl ester hydrochloride
PubChem CID 2756383
IUPAC Name methyl 4-aminobutanoate;hydrochloride
SMILES [Cl-].COC(=O)CCC[NH3+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC442911000
SDS View Documents
Thermo Scientific Chemicals
442911000
100 g Glass Bottle
Each for $821.99
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13031-60-2
153.61
WPGPRLVPWACBHW-UHFFFAOYSA-N
2756383
[Cl-].COC(=O)CCC[NH3+]
C5H12ClNO2
MFCD00043270
methyl 4-aminobutanoate hydrochloride, methyl 4-aminobutyrate hydrochloride, methyl 4-aminobutanoate hcl, methyl 4-aminobutyrate hcl, h-gamma-abu-ome.hcl, 4-aminobutyric acid methyl ester hydrochloride, 4-aminobutyric acid methyl ester, hydrochloride, 4-amino-n-butyric acid methyl ester hydrochloride, butanoic acid, 4-amino-, methyl ester, hydrochloride, 4-amino-butyric acid methyl ester hydrochloride
methyl 4-aminobutanoate;hydrochloride

Specifications

118°C to 119°C
98.5 to 101.5% (Argentometry)
Glass Bottle
methyl 4-aminobutanoate hydrochloride, methyl 4-aminobutyrate hydrochloride, methyl 4-aminobutanoate hcl, methyl 4-aminobutyrate hcl, h-gamma-abu-ome.hcl, 4-aminobutyric acid methyl ester hydrochloride, 4-aminobutyric acid methyl ester, hydrochloride, 4-amino-n-butyric acid methyl ester hydrochloride, butanoic acid, 4-amino-, methyl ester, hydrochloride, 4-amino-butyric acid methyl ester hydrochloride
[Cl-].COC(=O)CCC[NH3+]
100 g
2756383
99%
13031-60-2
C5H12ClNO2
MFCD00043270
WPGPRLVPWACBHW-UHFFFAOYSA-N
methyl 4-aminobutanoate;hydrochloride
153.61
153.61
Methyl 4-aminobutyrate hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only