missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-(Bromomethyl)-3-methoxybenzoate 97.0+%, TCI America™

$209.35 - $839.38

Chemical Identifiers

CAS 70264-94-7
Molecular Formula C10H11BrO3
Molecular Weight (g/mol) 259.099
MDL Number MFCD00270115
InChI Key UXSNXOMMJXTFEG-UHFFFAOYSA-N
Synonym 4-(Bromomethyl)-3-methoxybenzoic Acid Methyl Ester, Methyl 4-(Bromomethyl)-m-anisate, 4-(Bromomethyl)-m-anisic Acid Methyl Ester
PubChem CID 2759828
IUPAC Name methyl 4-(bromomethyl)-3-methoxybenzoate
SMILES COC1=C(C=CC(=C1)C(=O)OC)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M23565G
View Documents
TCI America
M23565G
5 g N/A
 
M235625G
View Documents
TCI America
M235625G
25 g N/A
 
Description

Description

Specifications

Chemical Identifiers

70264-94-7
259.099
UXSNXOMMJXTFEG-UHFFFAOYSA-N
2759828
COC1=C(C=CC(=C1)C(=O)OC)CBr
C10H11BrO3
MFCD00270115
4-(Bromomethyl)-3-methoxybenzoic Acid Methyl Ester, Methyl 4-(Bromomethyl)-m-anisate, 4-(Bromomethyl)-m-anisic Acid Methyl Ester
methyl 4-(bromomethyl)-3-methoxybenzoate

Specifications

70264-94-7
White-Yellow
MFCD00270115
3077
UXSNXOMMJXTFEG-UHFFFAOYSA-N
methyl 4-(bromomethyl)-3-methoxybenzoate
2759828
≥97.0% (GC)
Methyl 4-(Bromomethyl)-3-methoxybenzoate
93°C
C10H11BrO3
5 g
4-(Bromomethyl)-3-methoxybenzoic Acid Methyl Ester, Methyl 4-(Bromomethyl)-m-anisate, 4-(Bromomethyl)-m-anisic Acid Methyl Ester
COC1=C(C=CC(=C1)C(=O)OC)CBr
259.099
259.10
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'