Learn More
Methyl 4-bromopyridine-2-carboxylate, 97+%
CAS: 29681-42-3 | C7H6BrNO2 | 216.034 g/mol
$73.10 - $1041.53
Chemical Identifiers
| CAS | 29681-42-3 |
|---|---|
| Molecular Formula | C7H6BrNO2 |
| Molecular Weight (g/mol) | 216.034 |
| MDL Number | MFCD07374897 |
| InChI Key | JZFLATQBIPILFS-UHFFFAOYSA-N |
| PubChem CID | 12828947 |
| IUPAC Name | methyl 4-bromopyridine-2-carboxylate |
| SMILES | COC(=O)C1=NC=CC(=C1)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6432703
|
Thermo Scientific Chemicals
H6432703 |
1 g |
Each for $73.10
|
|
|||||
|
AAH6432706
|
Thermo Scientific Chemicals
H6432706 |
5 g |
Each for $250.67
|
|
|||||
|
AAH6432714
|
Thermo Scientific Chemicals
H6432714 |
25 g |
Each for $1,041.53
|
|
|||||
Description
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.
Solubility
Slightly soluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Chemical Identifiers
| 29681-42-3 | |
| 216.034 | |
| JZFLATQBIPILFS-UHFFFAOYSA-N | |
| methyl 4-bromopyridine-2-carboxylate |
| C7H6BrNO2 | |
| MFCD07374897 | |
| 12828947 | |
| COC(=O)C1=NC=CC(=C1)Br |
Specifications
| 29681-42-3 | |
| C7H6BrNO2 | |
| 1 g | |
| methyl 4-bromopicolinate, 4-bromo-pyridine-2-carboxylic acid methyl ester, methyl 4-bromo-2-pyridinecarboxylate, methyl4-bromopicolinate, 2-pyridinecarboxylic acid, 4-bromo-, methyl ester, 4-bromopyridine-2-carboxylic acid methyl ester, abbypharma ap-10-5912, pubchem12733, acmc-209hag, methyl 4-bromo-pyridine-2-carboxylate | |
| JZFLATQBIPILFS-UHFFFAOYSA-N | |
| methyl 4-bromopyridine-2-carboxylate | |
| 12828947 | |
| ≥97% | |
| Methyl 4-bromopyridine-2-carboxylate |
| 54°C to 56°C | |
| MFCD07374897 | |
| 473047 | |
| Slightly soluble in water. | |
| COC(=O)C1=NC=CC(=C1)Br | |
| 216.034 | |
| 216.03 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only