missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-chlorobutyrate, 98%, Thermo Scientific Chemicals

$59.81 - $124.90

Chemical Identifiers

CAS 3153-37-5
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.575
MDL Number MFCD00001003
InChI Key ZZUYIRISBMWFMV-UHFFFAOYSA-N
Synonym methyl 4-chlorobutyrate, butanoic acid, 4-chloro-, methyl ester, 4-chlorobutyric acid methyl ester, butyric acid, 4-chloro-, methyl ester, methyl .omega.-chlorobutyrate, 4-chlorobutanoic acid methyl ester, .gamma.-chlorobutyric acid methyl ester, methyl-4-chlorbutanoat, gamma-chlorobutyric acid methyl ester, methyl omega-chlorobutyrate
PubChem CID 76612
IUPAC Name methyl 4-chlorobutanoate
SMILES COC(=O)CCCCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0373414
View Documents
Thermo Scientific Chemicals
L0373414
25 g
Each for $59.81
 
AAL0373422
View Documents
Thermo Scientific Chemicals
L0373422
100 g
Each for $124.90
 
Description

Description

Methyl 4-chlorobutyrate is used as a pharmaceutical synthesis, industrial dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-chlorobutyrate is used as a pharmaceutical synthesis, industrial dye intermediates.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

3153-37-5
136.575
ZZUYIRISBMWFMV-UHFFFAOYSA-N
76612
COC(=O)CCCCl
C5H9ClO2
MFCD00001003
methyl 4-chlorobutyrate, butanoic acid, 4-chloro-, methyl ester, 4-chlorobutyric acid methyl ester, butyric acid, 4-chloro-, methyl ester, methyl .omega.-chlorobutyrate, 4-chlorobutanoic acid methyl ester, .gamma.-chlorobutyric acid methyl ester, methyl-4-chlorbutanoat, gamma-chlorobutyric acid methyl ester, methyl omega-chlorobutyrate
methyl 4-chlorobutanoate

Specifications

3153-37-5
175°C to 176°C
C5H9ClO2
MFCD00001003
UN3272
methyl 4-chlorobutyrate, butanoic acid, 4-chloro-, methyl ester, 4-chlorobutyric acid methyl ester, butyric acid, 4-chloro-, methyl ester, methyl .omega.-chlorobutyrate, 4-chlorobutanoic acid methyl ester, .gamma.-chlorobutyric acid methyl ester, methyl-4-chlorbutanoat, gamma-chlorobutyric acid methyl ester, methyl omega-chlorobutyrate
ZZUYIRISBMWFMV-UHFFFAOYSA-N
methyl 4-chlorobutanoate
76612
98%
1.125
59°C (138°F)
1.432
25 g
1745117
Slightly soluble in water.
COC(=O)CCCCl
136.575
136.58
Methyl 4-chlorobutyrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 221-592-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only