missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1462206

 View more versions of this product

Catalog No. AAL1462206

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate
122°C to 124°C
MFCD01075661
5258501
SEBIQMPNDQLYOX-UHFFFAOYSA-N
methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-ene-1-carboxylate
3766543
99%
39493-62-4
C9H12O4
5 g
methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-ene-1-carboxylate, methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-enecarboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-enecarboxylate, acmc-20akgd, methyl 4-hydroxy-6-methyl-2-oxo-cyclohex-3-en-1-oate, methyl 2-hydroxy-6-methyl-4-oxo-2-cyclohexene-1-carboxylate, 3-cyclohexene-1-carboxylicacid, 4-hydroxy-6-methyl-2-oxo-, methyl ester, 6-methyl-2-hydroxy-4-oxo-2-cyclohexene-1-carboxylic acid methyl ester
CC1CC(=O)C=C(C1C(=O)OC)O
184.191
184.19

Chemical Identifiers

39493-62-4
184.191
SEBIQMPNDQLYOX-UHFFFAOYSA-N
3766543
CC1CC(=O)C=C(C1C(=O)OC)O
C9H12O4
MFCD01075661
methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-ene-1-carboxylate, methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-enecarboxylate, methyl 4-hydroxy-6-methyl-2-oxocyclohex-3-enecarboxylate, acmc-20akgd, methyl 4-hydroxy-6-methyl-2-oxo-cyclohex-3-en-1-oate, methyl 2-hydroxy-6-methyl-4-oxo-2-cyclohexene-1-carboxylate, 3-cyclohexene-1-carboxylicacid, 4-hydroxy-6-methyl-2-oxo-, methyl ester, 6-methyl-2-hydroxy-4-oxo-2-cyclohexene-1-carboxylic acid methyl ester
methyl 2-hydroxy-6-methyl-4-oxocyclohex-2-ene-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only