missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-hydroxyphenylacetate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2253336

 View more versions of this product

Catalog No. AAB2253336


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl 4-hydroxyphenylacetate
55°C to 58°C
C9H10O3
500 g
methyl 4-hydroxyphenylacetate, methyl 2-4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methyl ester, benzeneacetic acid, 4-hydroxy-, methyl ester, methyl 4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methl ester, acetic acid, p-hydroxyphenyl-, methyl ester, p-hydroxyphenylacetic acid methyl ester, 4-hydroxybenzeneacetic acid methyl ester, 4-hydroxy-phenyl-acetic acid methyl ester
COC(=O)CC1=CC=C(C=C1)O
166.176
CHEBI:68078
≥98%
14199-15-6
163°C (5 mmHg)
MFCD00002387
2087538
XGDZEDRBLVIUMX-UHFFFAOYSA-N
methyl 2-(4-hydroxyphenyl)acetate
518900
166.18

Chemical Identifiers

14199-15-6
166.176
XGDZEDRBLVIUMX-UHFFFAOYSA-N
518900
methyl 2-(4-hydroxyphenyl)acetate
C9H10O3
MFCD00002387
methyl 4-hydroxyphenylacetate, methyl 2-4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methyl ester, benzeneacetic acid, 4-hydroxy-, methyl ester, methyl 4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methl ester, acetic acid, p-hydroxyphenyl-, methyl ester, p-hydroxyphenylacetic acid methyl ester, 4-hydroxybenzeneacetic acid methyl ester, 4-hydroxy-phenyl-acetic acid methyl ester
CHEBI:68078
COC(=O)CC1=CC=C(C=C1)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 238-050-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only