missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-nitrobenzoate, 99%, Thermo Scientific Chemicals

$105.00 - $352.45

Chemical Identifiers

CAS 619-50-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007350
InChI Key YOJAHJGBFDPSDI-UHFFFAOYSA-N
Synonym methyl p-nitrobenzoate, benzoic acid, 4-nitro-, methyl ester, p-nitrobenzoic acid methyl ester, methyl-p-nitrobenzoate, methyl-4-nitrobenzoate, benzoic acid, p-nitro-, methyl ester, p-carbomethoxynitrobenzene, unii-nmn8cth864, nmn8cth864, 4-nitrobenzoic acid methyl ester
PubChem CID 12083
IUPAC Name methyl 4-nitrobenzoate
SMILES COC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1292822
View Documents
Thermo Scientific Chemicals
A1292822
100 g
Each for $105.00
 
AAA1292836
View Documents
Thermo Scientific Chemicals
A1292836
500 g
Each for $352.45
 
Description

Description

It is employed in the biochemical characterization of NfsA, the Escherichia coli major nitroreductase exhibiting a high amino acid sequence homology to Frp, a Vibrio harveyi flavin oxidoreductase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the biochemical characterization of NfsA, the Escherichia coli major nitroreductase exhibiting a high amino acid sequence homology to Frp, a Vibrio harveyi flavin oxidoreductase.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

619-50-1
181.147
YOJAHJGBFDPSDI-UHFFFAOYSA-N
12083
COC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
C8H7NO4
MFCD00007350
methyl p-nitrobenzoate, benzoic acid, 4-nitro-, methyl ester, p-nitrobenzoic acid methyl ester, methyl-p-nitrobenzoate, methyl-4-nitrobenzoate, benzoic acid, p-nitro-, methyl ester, p-carbomethoxynitrobenzene, unii-nmn8cth864, nmn8cth864, 4-nitrobenzoic acid methyl ester
methyl 4-nitrobenzoate

Specifications

619-50-1
C8H7NO4
100 g
methyl p-nitrobenzoate, benzoic acid, 4-nitro-, methyl ester, p-nitrobenzoic acid methyl ester, methyl-p-nitrobenzoate, methyl-4-nitrobenzoate, benzoic acid, p-nitro-, methyl ester, p-carbomethoxynitrobenzene, unii-nmn8cth864, nmn8cth864, 4-nitrobenzoic acid methyl ester
COC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
181.147
181.15
Methyl 4-nitrobenzoate
95°C to 97°C
MFCD00007350
1912198
YOJAHJGBFDPSDI-UHFFFAOYSA-N
methyl 4-nitrobenzoate
12083
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-599-2

RTECSNumber : DH5775000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only