missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (4-Piperidyl)acetate Hydrochloride 98.0+%, TCI America™
$35.41 - $87.60
Chemical Identifiers
| CAS | 81270-37-3 |
|---|---|
| Molecular Formula | C8H16ClNO2 |
| Molecular Weight (g/mol) | 193.671 |
| InChI Key | ADBDFGZYGJGDNJ-UHFFFAOYSA-N |
| Synonym | (4-Piperidyl)acetic Acid Methyl Ester Hydrochloride |
| PubChem CID | 18423605 |
| IUPAC Name | methyl 2-piperidin-4-ylacetate;hydrochloride |
| SMILES | COC(=O)CC1CCNCC1.Cl |
Chemical Identifiers
| 81270-37-3 | |
| 193.671 | |
| (4-Piperidyl)acetic Acid Methyl Ester Hydrochloride | |
| methyl 2-piperidin-4-ylacetate;hydrochloride |
| C8H16ClNO2 | |
| ADBDFGZYGJGDNJ-UHFFFAOYSA-N | |
| 18423605 | |
| COC(=O)CC1CCNCC1.Cl |
Specifications
| 155°C | |
| White-Yellow | |
| (4-Piperidyl)acetic Acid Methyl Ester Hydrochloride | |
| COC(=O)CC1CCNCC1.Cl | |
| 1 g | |
| 18423605 | |
| ≥98.0% (T) | |
| Methyl (4-Piperidyl)acetate Hydrochloride |
| 81270-37-3 | |
| C8H16ClNO2 | |
| ADBDFGZYGJGDNJ-UHFFFAOYSA-N | |
| methyl 2-piperidin-4-ylacetate;hydrochloride | |
| 193.671 | |
| 193.67 | |
| Crystalline Powder |
Safety and Handling
TSCA : No