missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-Methoxy-3-oxovalerate 96.0+%, TCI America™
Supplier: TCI America M08125G
Specifications
| Methyl 5-Methoxy-3-oxovalerate | |
| Yellow | |
| C7H12O4 | |
| 5 g | |
| OPURNNHYTATPKM-UHFFFAOYSA-N | |
| methyl 5-methoxy-3-oxopentanoate | |
| 542416 | |
| ≥96.0% (GC) |
| 62462-05-9 | |
| 69°C | |
| MFCD00042840 | |
| 3-Keto-5-methoxyvaleric Acid Methyl Ester, 5-Methoxy-3-oxovaleric Acid Methyl Ester, Methyl 3-Keto-5-methoxyvalerate | |
| COCCC(=O)CC(=O)OC | |
| 160.17 | |
| 160.17 | |
| Liquid |
Chemical Identifiers
| 62462-05-9 | |
| 160.17 | |
| OPURNNHYTATPKM-UHFFFAOYSA-N | |
| 542416 | |
| COCCC(=O)CC(=O)OC |
| C7H12O4 | |
| MFCD00042840 | |
| 3-Keto-5-methoxyvaleric Acid Methyl Ester, 5-Methoxy-3-oxovaleric Acid Methyl Ester, Methyl 3-Keto-5-methoxyvalerate | |
| methyl 5-methoxy-3-oxopentanoate |
Safety and Handling
TSCA : No