missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 5-methylisoxazole-3-carboxylate, 97%, Thermo Scientific Chemicals

$215.49 - $642.81

Chemical Identifiers

CAS 19788-35-3
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.13
MDL Number MFCD00015895
InChI Key MVHHQOCEOUNTID-UHFFFAOYSA-N
Synonym methyl 5-methylisoxazole-3-carboxylate, methyl 5-methyl-3-isoxazolecarboxylate, 3-isoxazolecarboxylic acid, 5-methyl-, methyl ester, 5-methyl-3-isoxazolecarboxylic acid methyl ester, methyl 5-methyl-3-isoxazole carboxylate, methyl-5-methyl-3-isoxazole carboxylate, 5-methylisoxazole-3-carboxylic acid methyl ester, 5-methyl-isoxazole-3-carboxylic acid methyl ester, acmc-209xf5, ksc489s1r
PubChem CID 88245
IUPAC Name methyl 5-methyl-1,2-oxazole-3-carboxylate
SMILES COC(=O)C1=NOC(C)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1981414
View Documents
Thermo Scientific Chemicals
A1981414
25 g
Each for $215.49
 
AAA1981422
View Documents
Thermo Scientific Chemicals
A1981422
100 g
Each for $642.81
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19788-35-3
141.13
MVHHQOCEOUNTID-UHFFFAOYSA-N
88245
COC(=O)C1=NOC(C)=C1
C6H7NO3
MFCD00015895
methyl 5-methylisoxazole-3-carboxylate, methyl 5-methyl-3-isoxazolecarboxylate, 3-isoxazolecarboxylic acid, 5-methyl-, methyl ester, 5-methyl-3-isoxazolecarboxylic acid methyl ester, methyl 5-methyl-3-isoxazole carboxylate, methyl-5-methyl-3-isoxazole carboxylate, 5-methylisoxazole-3-carboxylic acid methyl ester, 5-methyl-isoxazole-3-carboxylic acid methyl ester, acmc-209xf5, ksc489s1r
methyl 5-methyl-1,2-oxazole-3-carboxylate

Specifications

19788-35-3
125°C (11 mmHg)
MFCD00015895
115704
MVHHQOCEOUNTID-UHFFFAOYSA-N
methyl 5-methyl-1,2-oxazole-3-carboxylate
88245
97%
91°C to 92°C
C6H7NO3
25 g
methyl 5-methylisoxazole-3-carboxylate, methyl 5-methyl-3-isoxazolecarboxylate, 3-isoxazolecarboxylic acid, 5-methyl-, methyl ester, 5-methyl-3-isoxazolecarboxylic acid methyl ester, methyl 5-methyl-3-isoxazole carboxylate, methyl-5-methyl-3-isoxazole carboxylate, 5-methylisoxazole-3-carboxylic acid methyl ester, 5-methyl-isoxazole-3-carboxylic acid methyl ester, acmc-209xf5, ksc489s1r
COC(=O)C1=NOC(C)=C1
141.13
141.13
Methyl 5-methylisoxazole-3-carboxylate
Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 243-311-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only